Assay Details
| Gene Symbol: TRGV9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC072396.1 | - | 1 - 2 | 345 | 72 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000390333.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000390334.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000390337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000390338.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000436911.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000436911.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000436911.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000436911.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000436911.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000443402.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000443402.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000443402.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000443402.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000443402.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000443402.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000443402.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000444775.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000444775.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000444775.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000444775.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000444775.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | NM_001003799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | NM_001003799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | NM_001003799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | NM_001003799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | NM_001003799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | NM_001003799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | NM_001003799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | NM_001003806.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | NM_001003806.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | NM_001003806.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | NM_001003806.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | NM_001003806.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | NM_001003806.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013538.hg.1 | NM_001003806.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000390333.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000390334.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000390337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000390338.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000436911.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000436911.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000436911.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000436911.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000436911.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000443402.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000443402.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000443402.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000443402.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000443402.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000443402.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000443402.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000444775.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000444775.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000444775.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000444775.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000444775.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000610889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | ENST00000619762.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | NM_001003799.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | NM_001003799.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | NM_001003799.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | NM_001003799.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | NM_001003799.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | NM_001003799.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | NM_001003799.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | NM_001003806.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | NM_001003806.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | NM_001003806.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | NM_001003806.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | NM_001003806.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | NM_001003806.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013538.hg.1 | NM_001003806.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 563421 | - | - |

TRGV9
Genomic Map