Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008537.hg.1 |
ENST00000576655.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008537.hg.1 |
ENST00000576655.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008537.hg.1 |
ENST00000576655.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008537.hg.1 |
ENST00000576655.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008537.hg.1 |
ENST00000576655.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008537.hg.1 |
ENST00000610896.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008537.hg.1 |
ENST00000610896.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008537.hg.1 |
ENST00000610896.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008537.hg.1 |
NM_001195256.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008537.hg.1 |
NM_001195256.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008537.hg.1 |
NM_001195256.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010179.hg.1 |
NM_001195256.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010179.hg.1 |
TC19000732.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010181.hg.1 |
NM_001195256.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010181.hg.1 |
TC19000732.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010180.hg.1 |
NM_001195256.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010180.hg.1 |
TC19000732.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900165940.hg.1 |
ENST00000576655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165940.hg.1 |
ENST00000610896.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165940.hg.1 |
NM_001195256.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900165940.hg.1 |
TC1900008537.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900165940.hg.1 |
ENST00000576655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165940.hg.1 |
ENST00000610896.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165940.hg.1 |
NM_001195256.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900165938.hg.1 |
ENST00000576655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165938.hg.1 |
ENST00000610896.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165938.hg.1 |
NM_001195256.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900165938.hg.1 |
TC1900008537.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900165938.hg.1 |
ENST00000576655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165938.hg.1 |
ENST00000610896.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165938.hg.1 |
NM_001195256.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008537.hg.1 |
ENST00000576655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008537.hg.1 |
ENST00000576655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008537.hg.1 |
ENST00000576655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008537.hg.1 |
ENST00000576655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008537.hg.1 |
ENST00000576655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008537.hg.1 |
ENST00000610896.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008537.hg.1 |
ENST00000610896.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008537.hg.1 |
ENST00000610896.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008537.hg.1 |
NM_001195256.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008537.hg.1 |
NM_001195256.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008537.hg.1 |
NM_001195256.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900165939.hg.1 |
ENST00000576655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165939.hg.1 |
ENST00000610896.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165939.hg.1 |
NM_001195256.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900165939.hg.1 |
TC1900008537.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900165939.hg.1 |
ENST00000576655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165939.hg.1 |
ENST00000610896.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165939.hg.1 |
NM_001195256.1 |
Refseq |