Assay Details
| Gene Symbol: ASPG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080464.2 | NP_001073933.2 | 12 - 13 | 1523 | 128 | |
| XM_005267590.4 | XP_005267647.1 | 12 - 13 | 1549 | 128 | ||
| XM_017021271.1 | XP_016876760.1 | 10 - 11 | 1990 | 128 | ||
| XM_017021272.1 | XP_016876761.1 | 10 - 11 | 1990 | 128 | ||
| XM_017021273.1 | XP_016876762.1 | 10 - 11 | 1990 | 128 | ||
| XM_017021275.1 | XP_016876764.1 | 10 - 11 | 1990 | 128 | ||
| XM_017021276.1 | XP_016876765.1 | 10 - 11 | 1990 | 128 | ||
| GenBank mRNA | AX781433.1 | - | 12 - 13 | 1530 | 128 | |
| AX781435.1 | - | 12 - 13 | 1438 | 128 | ||
| BC136637.1 | - | 12 - 13 | 1465 | 9020627 | 128 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14000811.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000546892.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000546892.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000546892.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000546892.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000546892.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000546892.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000546892.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000546892.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000546892.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000546892.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000546892.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000546892.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000546892.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000546892.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000546892.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000546892.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000546892.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000546892.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000546892.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000548372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000548372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000548372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000548372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000548372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000548372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000548372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000548372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000548372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000548372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000548372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000548372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000548372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000548372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000548372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000548372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000549809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000549809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000549809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000549809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000549809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000549809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000550583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000550583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000550583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000550583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000551170.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000551170.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000551170.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000551170.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000551871.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000551871.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000551871.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000551871.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000552126.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000552126.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000552126.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000552126.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000556267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | ENST00000556267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | NM_001080464.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | NM_001080464.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | NM_001080464.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | NM_001080464.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | NM_001080464.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | NM_001080464.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | NM_001080464.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | NM_001080464.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | NM_001080464.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | NM_001080464.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | NM_001080464.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | NM_001080464.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | NM_001080464.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | NM_001080464.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | NM_001080464.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | NM_001080464.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | NM_001080464.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | NM_001080464.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | NM_001080464.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | NM_001080464.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010696.hg.1 | NM_001080464.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14008842.hg.1 | ENST00000299234.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14008842.hg.1 | ENST00000455920.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14008842.hg.1 | ENST00000546892.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14008842.hg.1 | ENST00000551177.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14008842.hg.1 | NM_001080464.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14008842.hg.1 | TC14000811.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000546892.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000546892.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000546892.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000546892.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000546892.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000546892.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000546892.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000546892.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000546892.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000546892.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000546892.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000546892.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000546892.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000546892.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000546892.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000546892.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000546892.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000546892.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000546892.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000548372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000548372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000548372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000548372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000548372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000548372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000548372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000548372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000548372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000548372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000548372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000548372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000548372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000548372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000548372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000548372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000549809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000549809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000549809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000549809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000549809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000549809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000550583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000550583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000550583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000550583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000551170.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000551170.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000551170.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000551170.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000551871.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000551871.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000551871.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000551871.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000552126.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000552126.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000552126.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000552126.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000556267.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | ENST00000556267.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | NM_001080464.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | NM_001080464.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | NM_001080464.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | NM_001080464.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | NM_001080464.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | NM_001080464.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | NM_001080464.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | NM_001080464.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | NM_001080464.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | NM_001080464.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | NM_001080464.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | NM_001080464.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | NM_001080464.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | NM_001080464.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | NM_001080464.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | NM_001080464.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | NM_001080464.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | NM_001080464.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | NM_001080464.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | NM_001080464.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010696.hg.1 | NM_001080464.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400180938.hg.1 | ENST00000546892.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180938.hg.1 | ENST00000550583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180938.hg.1 | NM_001080464.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400180938.hg.1 | ENST00000546892.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180938.hg.1 | ENST00000550583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180938.hg.1 | NM_001080464.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400180938.hg.1 | TC1400010696.hg.1 | Clariom D TC |

ASPG
Genomic Map