| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11003252.hg.1 |
ENST00000501599.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11003252.hg.1 |
ENST00000511927.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11003252.hg.1 |
NR_038360.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11003252.hg.1 |
TC11000245.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007010.hg.1 |
ENST00000501599.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007010.hg.1 |
ENST00000501599.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007010.hg.1 |
ENST00000501599.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007010.hg.1 |
ENST00000501599.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007010.hg.1 |
ENST00000501599.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007010.hg.1 |
ENST00000501599.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007010.hg.1 |
ENST00000511927.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007010.hg.1 |
ENST00000511927.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007010.hg.1 |
ENST00000511927.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007010.hg.1 |
ENST00000542172.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007010.hg.1 |
ENST00000542172.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007010.hg.1 |
ENST00000542172.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007010.hg.1 |
NR_038360.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007010.hg.1 |
NR_038360.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007010.hg.1 |
NR_038360.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007010.hg.1 |
NR_038360.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007010.hg.1 |
NR_038360.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007010.hg.1 |
NR_038360.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11003253.hg.1 |
ENST00000501599.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11003253.hg.1 |
ENST00000511927.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11003253.hg.1 |
NR_038360.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11003253.hg.1 |
TC11000245.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1100149410.hg.1 |
ENST00000501599.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100149410.hg.1 |
ENST00000511927.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100149410.hg.1 |
NR_038360.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100149410.hg.1 |
ENST00000501599.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100149410.hg.1 |
ENST00000511927.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100149410.hg.1 |
NR_038360.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100149410.hg.1 |
TC1100007010.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100149409.hg.1 |
ENST00000501599.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100149409.hg.1 |
ENST00000511927.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100149409.hg.1 |
NR_038360.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100149409.hg.1 |
ENST00000501599.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100149409.hg.1 |
ENST00000511927.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100149409.hg.1 |
NR_038360.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100149409.hg.1 |
TC1100007010.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1100007010.hg.1 |
ENST00000501599.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007010.hg.1 |
ENST00000501599.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007010.hg.1 |
ENST00000501599.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007010.hg.1 |
ENST00000501599.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007010.hg.1 |
ENST00000501599.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007010.hg.1 |
ENST00000501599.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007010.hg.1 |
ENST00000511927.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007010.hg.1 |
ENST00000511927.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007010.hg.1 |
ENST00000511927.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007010.hg.1 |
ENST00000542172.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007010.hg.1 |
ENST00000542172.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007010.hg.1 |
ENST00000542172.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007010.hg.1 |
NR_038360.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007010.hg.1 |
NR_038360.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007010.hg.1 |
NR_038360.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007010.hg.1 |
NR_038360.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007010.hg.1 |
NR_038360.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007010.hg.1 |
NR_038360.1 |
Refseq |