| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012375.hg.1 |
ENST00000438797.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012375.hg.1 |
ENST00000438797.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012375.hg.1 |
NR_131786.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012375.hg.1 |
NR_131786.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012375.hg.1 |
NR_131786.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012375.hg.1 |
NR_131786.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600185296.hg.1 |
ENST00000438797.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600185296.hg.1 |
NR_131786.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600185296.hg.1 |
ENST00000438797.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600185296.hg.1 |
NR_131786.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600185296.hg.1 |
TC0600012375.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600185297.hg.1 |
ENST00000438797.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600185297.hg.1 |
NR_131786.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600185297.hg.1 |
ENST00000438797.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600185297.hg.1 |
NR_131786.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600185297.hg.1 |
TC0600012375.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0600012375.hg.1 |
ENST00000438797.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012375.hg.1 |
ENST00000438797.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012375.hg.1 |
NR_131786.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012375.hg.1 |
NR_131786.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012375.hg.1 |
NR_131786.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012375.hg.1 |
NR_131786.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600185302.hg.1 |
NR_131786.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600185302.hg.1 |
NR_131786.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600185302.hg.1 |
TC0600012375.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600185300.hg.1 |
NR_131786.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600185300.hg.1 |
NR_131786.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600185300.hg.1 |
TC0600012375.hg.1 |
Clariom D TC |