Assay Details
| Gene Symbol: FAM71E2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145402.1 | NP_001138874.1 | 9 - 10 | 2909 | 105 | |
| GenBank mRNA | AK127979.1 | - | 8 - 9 | 2890 | 105 | |
| AL834316.1 | - | 2 - 3 | 1667 | 105 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001869.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19025448.hg.1 | ENST00000424985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19025448.hg.1 | NM_001145402.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19025448.hg.1 | TC19001869.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000424985.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | ENST00000585734.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012052.hg.1 | NM_001145402.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900208359.hg.1 | ENST00000424985.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900208359.hg.1 | ENST00000585734.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900208359.hg.1 | NM_001145402.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900208359.hg.1 | TC1900012052.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900208359.hg.1 | ENST00000424985.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900208359.hg.1 | ENST00000585734.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900208359.hg.1 | NM_001145402.1 | Refseq |

FAM71E2
Genomic Map