Assay Details
| Gene Symbol: LOC101929460 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_134622.1 | 1 - 2 | 72 | 100 | ||
| GenBank mRNA | BC040917.1 | - | 1 - 2 | 72 | 100 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105378146 | LOC105378146 | -69.8kb | Sense | Chr. 6 - 168927741 - 168964382 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC102724357 | LOC102724357 | 32.1kb | Anti-sense | Chr. 6 - 169067786 - 169069424 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC101929460
Genomic Map