| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010177.hg.1 |
ENST00000444219.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010177.hg.1 |
ENST00000444219.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010177.hg.1 |
ENST00000444219.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010177.hg.1 |
ENST00000444219.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010177.hg.1 |
ENST00000444219.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010177.hg.1 |
ENST00000444219.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010177.hg.1 |
ENST00000444219.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010177.hg.1 |
ENST00000444219.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010177.hg.1 |
NR_125862.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010177.hg.1 |
NR_125862.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010177.hg.1 |
NR_125862.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010177.hg.1 |
NR_125862.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010177.hg.1 |
NR_125862.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010177.hg.1 |
NR_125862.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600010177.hg.1 |
ENST00000444219.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600010177.hg.1 |
ENST00000444219.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600010177.hg.1 |
ENST00000444219.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600010177.hg.1 |
ENST00000444219.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600010177.hg.1 |
ENST00000444219.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600010177.hg.1 |
ENST00000444219.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600010177.hg.1 |
ENST00000444219.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600010177.hg.1 |
ENST00000444219.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600010177.hg.1 |
NR_125862.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600010177.hg.1 |
NR_125862.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600010177.hg.1 |
NR_125862.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600010177.hg.1 |
NR_125862.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600010177.hg.1 |
NR_125862.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600010177.hg.1 |
NR_125862.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600169534.hg.1 |
ENST00000444219.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600169534.hg.1 |
NR_125862.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600169534.hg.1 |
ENST00000444219.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600169534.hg.1 |
NR_125862.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600169534.hg.1 |
TC0600010177.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600169535.hg.1 |
ENST00000444219.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600169535.hg.1 |
NR_125862.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600169535.hg.1 |
ENST00000444219.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600169535.hg.1 |
NR_125862.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600169535.hg.1 |
TC0600010177.hg.1 |
Clariom D TC |