| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008520.hg.1 |
ENST00000548011.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008520.hg.1 |
ENST00000552554.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008520.hg.1 |
ENST00000552554.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008520.hg.1 |
ENST00000552554.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008520.hg.1 |
ENST00000552554.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008520.hg.1 |
ENST00000552554.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008520.hg.1 |
ENST00000552554.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008520.hg.1 |
ENST00000552554.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008520.hg.1 |
ENST00000626376.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008520.hg.1 |
ENST00000626376.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008520.hg.1 |
NR_077225.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008520.hg.1 |
NR_077225.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008520.hg.1 |
NR_077225.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008520.hg.1 |
NR_077225.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008520.hg.1 |
NR_077225.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008520.hg.1 |
NR_077225.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008520.hg.1 |
NR_077225.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200162452.hg.1 |
ENST00000552554.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200162452.hg.1 |
NR_077225.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200162452.hg.1 |
ENST00000552554.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200162452.hg.1 |
NR_077225.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200162452.hg.1 |
TC1200008520.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200162456.hg.1 |
ENST00000552554.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200162456.hg.1 |
NR_077225.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200162456.hg.1 |
ENST00000552554.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200162456.hg.1 |
NR_077225.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200162456.hg.1 |
TC1200008520.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1200008520.hg.1 |
ENST00000548011.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008520.hg.1 |
ENST00000552554.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008520.hg.1 |
ENST00000552554.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008520.hg.1 |
ENST00000552554.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008520.hg.1 |
ENST00000552554.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008520.hg.1 |
ENST00000552554.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008520.hg.1 |
ENST00000552554.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008520.hg.1 |
ENST00000552554.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008520.hg.1 |
ENST00000626376.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008520.hg.1 |
ENST00000626376.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008520.hg.1 |
NR_077225.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008520.hg.1 |
NR_077225.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008520.hg.1 |
NR_077225.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008520.hg.1 |
NR_077225.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008520.hg.1 |
NR_077225.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008520.hg.1 |
NR_077225.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008520.hg.1 |
NR_077225.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200162453.hg.1 |
ENST00000552554.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200162453.hg.1 |
ENST00000626376.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200162453.hg.1 |
NR_077225.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200162453.hg.1 |
ENST00000552554.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200162453.hg.1 |
ENST00000626376.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200162453.hg.1 |
NR_077225.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200162453.hg.1 |
TC1200008520.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200162455.hg.1 |
ENST00000552554.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200162455.hg.1 |
NR_077225.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200162455.hg.1 |
ENST00000552554.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200162455.hg.1 |
NR_077225.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200162455.hg.1 |
TC1200008520.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200162458.hg.1 |
ENST00000552554.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200162458.hg.1 |
NR_077225.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200162458.hg.1 |
ENST00000552554.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200162458.hg.1 |
NR_077225.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200162458.hg.1 |
TC1200008520.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200162457.hg.1 |
ENST00000548011.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200162457.hg.1 |
ENST00000552554.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200162457.hg.1 |
NR_077225.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200162457.hg.1 |
ENST00000548011.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200162457.hg.1 |
ENST00000552554.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200162457.hg.1 |
NR_077225.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200162457.hg.1 |
TC1200008520.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200162454.hg.1 |
ENST00000552554.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200162454.hg.1 |
NR_077225.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200162454.hg.1 |
ENST00000552554.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200162454.hg.1 |
NR_077225.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200162454.hg.1 |
TC1200008520.hg.1 |
Clariom D TC |