| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392977.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392977.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392977.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392977.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392977.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392977.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392977.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392977.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392977.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392977.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392977.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392977.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392977.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392977.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392977.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392977.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392977.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392977.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392977.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392977.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392977.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392977.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392977.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392977.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392977.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392977.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392977.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392977.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392977.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392977.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392977.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392979.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392979.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392979.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392979.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392979.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392979.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392979.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392979.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392979.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392979.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392979.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392979.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392979.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392979.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392979.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392979.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392979.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392979.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392979.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392979.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392979.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392979.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392979.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392979.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392979.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392979.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392979.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392979.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392979.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392979.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392979.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000392979.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000546991.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000546991.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000546991.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000546991.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000546991.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000546991.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000546991.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000548940.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000548940.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000548940.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000548940.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000549155.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000549155.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000549155.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000549155.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000549155.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000549155.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000549155.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000549155.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000549155.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000549155.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000549155.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000549155.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000549234.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000549234.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000549234.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000549234.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000549234.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000549234.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000550015.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000550015.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000550015.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000550015.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000550015.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000550015.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000550015.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000550015.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000550015.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000550015.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000550015.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000550015.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000550015.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000550015.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000550015.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000550015.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000550015.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000551148.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000551148.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000551148.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000551148.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
ENST00000551148.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
NM_178826.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
NM_178826.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
NM_178826.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
NM_178826.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
NM_178826.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
NM_178826.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
NM_178826.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
NM_178826.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
NM_178826.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
NM_178826.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
NM_178826.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
NM_178826.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
NM_178826.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
NM_178826.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
NM_178826.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
NM_178826.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
NM_178826.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
NM_178826.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
NM_178826.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
NM_178826.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
NM_178826.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
NM_178826.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
NM_178826.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
NM_178826.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
NM_178826.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
NM_178826.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
NM_178826.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
NM_178826.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
NM_178826.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
NM_178826.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
NM_178826.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
NM_178826.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
NM_178826.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008601.hg.1 |
NM_178826.3 |
Refseq |