Assay Details
| Gene Symbol: DCAF13P3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027642.1 | 1 | 522 | 79 | ||
| GenBank mRNA | DB137411.1 | - | 1 | 357 | 79 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15000400.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3420426 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 18819 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3420426 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3420426 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143837 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143830 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143819 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143818 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09651164 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143779 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143785 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143794 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143826 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143822 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143834 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143787 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09651121 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143800 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143797 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143808 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09651112 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143799 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143840 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143809 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143804 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143811 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09651131 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143792 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143795 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143810 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143814 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09651116 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143817 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09651151 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143777 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143793 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143807 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09651158 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09651162 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143780 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143801 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143788 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09651117 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143832 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09651124 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143806 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09651171 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143828 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143827 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09651119 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143816 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09651117 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09651116 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143834 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143822 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143806 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143832 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143788 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143799 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09651112 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143801 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143828 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143830 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143785 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09651124 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143816 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143837 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143819 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143810 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143795 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143807 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143779 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09651121 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09651164 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09651171 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143840 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143800 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143808 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143814 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143818 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143777 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143792 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09651131 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143794 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143797 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143804 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09651162 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143817 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09651151 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143809 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143827 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143826 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143780 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143811 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143787 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09651119 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30143793 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09651158 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09651156 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09651110 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09651147 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09651108 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09651126 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09651173 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09651154 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09651159 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09651123 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09651145 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09651135 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09651129 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09651104 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09651132 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09651113 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09651137 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09651151 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09651171 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09651142 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09651127 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09651099 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09651165 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09651140 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09651162 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09651120 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09651168 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09651160 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09651118 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09651122 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09651115 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09651124 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09651153 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09651106 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09651101 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09651149 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR15003751.hg.1 | NR_027642.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15003751.hg.1 | TC15000400.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007218.hg.1 | ENST00000561147.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007218.hg.1 | NR_027642.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007218.hg.1 | NR_027642.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007218.hg.1 | NR_027642.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500151514.hg.1 | NR_027642.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500151514.hg.1 | NR_027642.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500151514.hg.1 | TC1500007218.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500151516.hg.1 | NR_027642.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500151516.hg.1 | NR_027642.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500151516.hg.1 | TC1500007218.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500151515.hg.1 | ENST00000561147.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500151515.hg.1 | NR_027642.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500151515.hg.1 | ENST00000561147.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500151515.hg.1 | NR_027642.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500151515.hg.1 | TC1500007218.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1500007218.hg.1 | ENST00000561147.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007218.hg.1 | NR_027642.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007218.hg.1 | NR_027642.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007218.hg.1 | NR_027642.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A052892 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A052892 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3200465 | NR_027642.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3291501 | NR_027642.1 | human_transcriptome |

DCAF13P3
Genomic Map