Assay Details
| Gene Symbol: SOCS2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK054607.1 | - | 1 - 2 | 200 | 88 | |
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3375002 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P333413 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 1559315_PM_s_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000547845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000547845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000547845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000547845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000547845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000547845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000547845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000547845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000551626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000551626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000551626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000551626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000551626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | ENST00000551626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | NR_038263.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | NR_038263.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | NR_038263.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | NR_038263.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011541.hg.1 | NR_038263.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000499137.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000500986.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000547845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000547845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000547845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000547845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000547845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000547845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000547845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000547845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000551626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000551626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000551626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000551626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000551626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | ENST00000551626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | NR_038263.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | NR_038263.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | NR_038263.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | NR_038263.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011541.hg.1 | NR_038263.1 | Refseq |
| Human Genome U133 Plus 2.0 Array | 1559315_PM_s_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1559315_s_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559315_s_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559315_s_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559315_s_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559315_s_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559315_s_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559315_s_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559315_s_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559315_s_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559315_s_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559315_s_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559315_s_at4 | - | - |
| Human Genome U95 Set | 64834_PM_at | - | - |
| Human Genome U95 Set | 64834_at | - | - |
| Human Genome U95 Set | 73954_PM_at | - | - |
| Human Genome U95 Set | 73954_at | - | - |
| Human Genome U95 Set | 70162_at | - | - |
| Human Genome U95 Set | 70162_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A047893 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A047893 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1699188 | AK054607.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1699188 | AK054607.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1699188 | AK054607.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1699188 | AK054607.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1699188 | AK054607.1 | human_transcriptome |

SOCS2-AS1
Genomic Map