Assay Details
| Gene Symbol: LINC00598 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024505.1 | 6 - 7 | 1818 | 127 | ||
| GenBank mRNA | AY116214.1 | - | 6 - 7 | 1816 | 127 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 548 | LINC00548 | -230.4kb | Sense | Chr. 13 - 40194509 - 40220502 on NCBI Build GRCh38 | |
| Downstream | forkhead box O1 | FOXO1 |
FKH1, FKHR, FOXO1A |
97.0kb | Sense | Chr. 13 - 40555664 - 40666597 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC13001718.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | ENST00000400430.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | ENST00000400430.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | ENST00000400430.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | ENST00000400430.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | ENST00000400430.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | ENST00000400430.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | ENST00000400430.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | ENST00000400430.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | ENST00000400430.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | ENST00000400430.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | ENST00000400430.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | ENST00000400430.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | ENST00000400431.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | ENST00000400431.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | ENST00000542632.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | ENST00000542632.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | ENST00000542632.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | ENST00000542632.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | ENST00000542632.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | ENST00000542632.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | ENST00000542632.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | ENST00000542632.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | ENST00000542632.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | ENST00000542632.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | ENST00000542632.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | ENST00000542632.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | ENST00000542632.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | ENST00000615137.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | ENST00000615137.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | ENST00000615137.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | ENST00000615137.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | ENST00000616706.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | ENST00000616706.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | ENST00000616706.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | ENST00000616706.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | ENST00000616706.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | ENST00000616706.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | ENST00000616706.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | ENST00000616706.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | ENST00000616706.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | ENST00000616706.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | ENST00000616706.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | NR_024505.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | NR_024505.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | NR_024505.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | NR_024505.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | NR_024505.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | NR_024505.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | NR_024505.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | NR_024505.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | NR_024505.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | NR_024505.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | NR_024505.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | NR_024506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | NR_024506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | NR_024506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | NR_024506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | NR_024506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | NR_024506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | NR_024506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | NR_024506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | NR_024507.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | NR_024507.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | NR_024507.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | NR_024507.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | NR_024507.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | NR_024507.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | NR_024507.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | NR_024507.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010036.hg.1 | NR_024507.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13006751.hg.1 | NR_024505.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13006751.hg.1 | TC13001718.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR13006752.hg.1 | ENST00000400430.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13006752.hg.1 | NR_024505.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13006752.hg.1 | TC13001718.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR13006753.hg.1 | ENST00000400430.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13006753.hg.1 | NR_024505.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13006753.hg.1 | NR_024507.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13006753.hg.1 | TC13001718.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1300167658.hg.1 | ENST00000400430.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300167658.hg.1 | ENST00000400430.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300167658.hg.1 | ENST00000542632.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300167658.hg.1 | ENST00000616706.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300167658.hg.1 | NR_024505.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300167658.hg.1 | ENST00000400430.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300167658.hg.1 | ENST00000400430.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300167658.hg.1 | ENST00000542632.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300167658.hg.1 | ENST00000616706.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300167658.hg.1 | NR_024505.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300167658.hg.1 | TC1300010036.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1300167659.hg.1 | ENST00000400430.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300167659.hg.1 | ENST00000400430.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300167659.hg.1 | ENST00000542632.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300167659.hg.1 | ENST00000616706.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300167659.hg.1 | NR_024505.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300167659.hg.1 | NR_024507.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300167659.hg.1 | ENST00000400430.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300167659.hg.1 | ENST00000400430.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300167659.hg.1 | ENST00000542632.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300167659.hg.1 | ENST00000616706.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300167659.hg.1 | NR_024505.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300167659.hg.1 | NR_024507.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300167659.hg.1 | TC1300010036.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | ENST00000400430.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | ENST00000400430.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | ENST00000400430.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | ENST00000400430.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | ENST00000400430.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | ENST00000400430.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | ENST00000400430.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | ENST00000400430.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | ENST00000400430.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | ENST00000400430.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | ENST00000400430.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | ENST00000400430.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | ENST00000400431.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | ENST00000400431.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | ENST00000542632.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | ENST00000542632.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | ENST00000542632.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | ENST00000542632.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | ENST00000542632.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | ENST00000542632.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | ENST00000542632.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | ENST00000542632.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | ENST00000542632.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | ENST00000542632.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | ENST00000542632.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | ENST00000542632.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | ENST00000542632.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | ENST00000615137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | ENST00000615137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | ENST00000615137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | ENST00000615137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | ENST00000616706.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | ENST00000616706.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | ENST00000616706.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | ENST00000616706.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | ENST00000616706.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | ENST00000616706.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | ENST00000616706.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | ENST00000616706.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | ENST00000616706.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | ENST00000616706.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | ENST00000616706.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | NR_024505.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | NR_024505.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | NR_024505.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | NR_024505.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | NR_024505.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | NR_024505.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | NR_024505.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | NR_024505.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | NR_024505.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | NR_024505.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | NR_024505.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | NR_024506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | NR_024506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | NR_024506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | NR_024506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | NR_024506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | NR_024506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | NR_024506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | NR_024506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | NR_024507.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | NR_024507.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | NR_024507.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | NR_024507.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | NR_024507.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | NR_024507.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | NR_024507.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | NR_024507.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010036.hg.1 | NR_024507.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300167657.hg.1 | ENST00000542632.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300167657.hg.1 | ENST00000616706.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300167657.hg.1 | NR_024505.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300167657.hg.1 | ENST00000542632.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300167657.hg.1 | ENST00000616706.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300167657.hg.1 | NR_024505.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300167657.hg.1 | TC1300010036.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 462606 | - | - |

LINC00598
Genomic Map