Assay Details
| Gene Symbol: SLC38A8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080442.2 | NP_001073911.1 | 2 - 3 | 388 | 99 | |
| XM_017022946.1 | XP_016878435.1 | 4 - 5 | 853 | 99 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16001305.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600011043.hg.1 | ENST00000299709.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011043.hg.1 | ENST00000299709.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011043.hg.1 | ENST00000299709.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011043.hg.1 | ENST00000299709.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011043.hg.1 | ENST00000299709.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011043.hg.1 | ENST00000299709.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011043.hg.1 | ENST00000299709.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011043.hg.1 | ENST00000299709.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011043.hg.1 | ENST00000299709.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011043.hg.1 | ENST00000299709.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011043.hg.1 | ENST00000299709.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011043.hg.1 | ENST00000568003.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011043.hg.1 | ENST00000568003.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011043.hg.1 | ENST00000568003.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011043.hg.1 | ENST00000568003.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011043.hg.1 | ENST00000568003.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011043.hg.1 | ENST00000568178.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011043.hg.1 | ENST00000568178.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011043.hg.1 | ENST00000568178.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011043.hg.1 | ENST00000568178.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011043.hg.1 | ENST00000568178.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011043.hg.1 | ENST00000568178.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011043.hg.1 | ENST00000568178.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011043.hg.1 | ENST00000568178.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011043.hg.1 | ENST00000569816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011043.hg.1 | ENST00000569816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011043.hg.1 | ENST00000569816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011043.hg.1 | NM_001080442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011043.hg.1 | NM_001080442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011043.hg.1 | NM_001080442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011043.hg.1 | NM_001080442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011043.hg.1 | NM_001080442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011043.hg.1 | NM_001080442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011043.hg.1 | NM_001080442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011043.hg.1 | NM_001080442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011043.hg.1 | NM_001080442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011043.hg.1 | NM_001080442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011043.hg.1 | NM_001080442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011043.hg.1 | NM_001080442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018096.hg.1 | ENST00000299709.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018096.hg.1 | NM_001080442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018096.hg.1 | TC16001305.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018097.hg.1 | ENST00000299709.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018097.hg.1 | NM_001080442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018097.hg.1 | TC16001305.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018098.hg.1 | ENST00000299709.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018098.hg.1 | NM_001080442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018098.hg.1 | TC16001305.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1600011043.hg.1 | ENST00000299709.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011043.hg.1 | ENST00000299709.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011043.hg.1 | ENST00000299709.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011043.hg.1 | ENST00000299709.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011043.hg.1 | ENST00000299709.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011043.hg.1 | ENST00000299709.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011043.hg.1 | ENST00000299709.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011043.hg.1 | ENST00000299709.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011043.hg.1 | ENST00000299709.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011043.hg.1 | ENST00000299709.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011043.hg.1 | ENST00000299709.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011043.hg.1 | ENST00000568003.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011043.hg.1 | ENST00000568003.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011043.hg.1 | ENST00000568003.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011043.hg.1 | ENST00000568003.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011043.hg.1 | ENST00000568003.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011043.hg.1 | ENST00000568178.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011043.hg.1 | ENST00000568178.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011043.hg.1 | ENST00000568178.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011043.hg.1 | ENST00000568178.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011043.hg.1 | ENST00000568178.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011043.hg.1 | ENST00000568178.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011043.hg.1 | ENST00000568178.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011043.hg.1 | ENST00000568178.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011043.hg.1 | ENST00000569816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011043.hg.1 | ENST00000569816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011043.hg.1 | ENST00000569816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011043.hg.1 | NM_001080442.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011043.hg.1 | NM_001080442.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011043.hg.1 | NM_001080442.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011043.hg.1 | NM_001080442.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011043.hg.1 | NM_001080442.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011043.hg.1 | NM_001080442.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011043.hg.1 | NM_001080442.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011043.hg.1 | NM_001080442.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011043.hg.1 | NM_001080442.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011043.hg.1 | NM_001080442.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011043.hg.1 | NM_001080442.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011043.hg.1 | NM_001080442.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600185731.hg.1 | ENST00000299709.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185731.hg.1 | ENST00000568178.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185731.hg.1 | NM_001080442.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600185731.hg.1 | TC1600011043.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600185731.hg.1 | ENST00000299709.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185731.hg.1 | ENST00000568178.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185731.hg.1 | NM_001080442.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600185729.hg.1 | ENST00000299709.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185729.hg.1 | ENST00000568178.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185729.hg.1 | ENST00000569816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185729.hg.1 | NM_001080442.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600185729.hg.1 | TC1600011043.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600185729.hg.1 | ENST00000299709.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185729.hg.1 | ENST00000568178.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185729.hg.1 | ENST00000569816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185729.hg.1 | NM_001080442.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600185728.hg.1 | ENST00000299709.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185728.hg.1 | ENST00000568178.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185728.hg.1 | ENST00000569816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185728.hg.1 | NM_001080442.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600185728.hg.1 | TC1600011043.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600185728.hg.1 | ENST00000299709.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185728.hg.1 | ENST00000568178.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185728.hg.1 | ENST00000569816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185728.hg.1 | NM_001080442.2 | Refseq |

SLC38A8
Genomic Map