| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Clariom D Assay |
PSR1700200300.hg.1 |
ENST00000570890.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700200300.hg.1 |
ENST00000570921.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700200300.hg.1 |
ENST00000572215.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700200300.hg.1 |
NR_002710.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700200300.hg.1 |
NR_120453.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700200300.hg.1 |
TC1700012179.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700200300.hg.1 |
ENST00000570890.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700200300.hg.1 |
ENST00000570921.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700200300.hg.1 |
ENST00000572215.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700200300.hg.1 |
NR_002710.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700200300.hg.1 |
NR_120453.1 |
Refseq |