Assay Details
| Gene Symbol: ALOX12P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120453.1 | 8 - 9 | 1169 | 75 | ||
| GenBank mRNA | BC041851.1 | - | 11 - 12 | 1903 | 75 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | tektin 1 | TEKT1 | -66.5kb | Anti-sense | Chr. 17 - 6799981 - 6831741 on NCBI Build GRCh38 | |
| Downstream | ALOX12 antisense RNA 1 | ALOX12-AS1 | 86.4kb | Anti-sense | Chr. 17 - 6985123 - 7012334 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

ALOX12P2
Genomic Map