Assay Details
| Gene Symbol: IGSF23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205280.1 | NP_001192209.1 | 1 - 2 | 137 | 68 | |
| XM_017026318.1 | XP_016881807.1 | 1 - 2 | 1190 | 68 | ||
| GenBank mRNA | BX112065.1 | - | 1 - 2 | 137 | 68 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000623.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19008638.hg.1 | ENST00000402988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008638.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008638.hg.1 | TC19000623.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008639.hg.1 | ENST00000402988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008639.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008639.hg.1 | TC19000623.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008634.hg.1 | ENST00000402988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008634.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008634.hg.1 | TC19000623.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000402988.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000402988.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000402988.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000402988.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000402988.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000402988.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000402988.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000402988.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000402988.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000428245.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000428245.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000428245.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000428245.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000428245.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000428245.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000428245.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000428245.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000428245.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000441389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000441389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000441389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000441389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000441389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000441389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000592507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000592507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000592507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | ENST00000592507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008271.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900163130.hg.1 | ENST00000402988.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163130.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900163130.hg.1 | TC1900008271.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900163130.hg.1 | ENST00000402988.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163130.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000402988.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000402988.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000402988.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000402988.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000402988.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000402988.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000402988.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000402988.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000402988.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000428245.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000428245.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000428245.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000428245.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000428245.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000428245.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000428245.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000428245.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000428245.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000441389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000441389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000441389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000441389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000441389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000441389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000592507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000592507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000592507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | ENST00000592507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008271.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900163137.hg.1 | ENST00000402988.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163137.hg.1 | ENST00000428245.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163137.hg.1 | ENST00000592507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163137.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900163137.hg.1 | TC1900008271.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900163137.hg.1 | ENST00000402988.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163137.hg.1 | ENST00000428245.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163137.hg.1 | ENST00000592507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163137.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900163138.hg.1 | ENST00000402988.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163138.hg.1 | ENST00000428245.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163138.hg.1 | ENST00000441389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163138.hg.1 | ENST00000592507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163138.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900163138.hg.1 | TC1900008271.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900163138.hg.1 | ENST00000402988.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163138.hg.1 | ENST00000428245.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163138.hg.1 | ENST00000441389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163138.hg.1 | ENST00000592507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163138.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900163139.hg.1 | ENST00000402988.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163139.hg.1 | ENST00000428245.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163139.hg.1 | ENST00000441389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163139.hg.1 | NM_001205280.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900163139.hg.1 | TC1900008271.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900163139.hg.1 | ENST00000402988.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163139.hg.1 | ENST00000428245.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163139.hg.1 | ENST00000441389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163139.hg.1 | NM_001205280.1 | Refseq |

IGSF23
Genomic Map