Assay Details
| Gene Symbol: GPR19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006143.2 | NP_006134.1 | 2 - 3 | 175 | 87 | |
| XM_011520623.2 | XP_011518925.1 | 2 - 3 | 647 | 87 | ||
| XM_011520624.2 | XP_011518926.1 | 2 - 3 | 301 | 87 | ||
| XM_011520625.2 | XP_011518927.1 | 2 - 3 | 1159 | 87 | ||
| XM_017019212.1 | XP_016874701.1 | 2 - 3 | 1159 | 87 | ||
| XM_017019213.1 | XP_016874702.1 | 2 - 3 | 1355 | 87 | ||
| XM_017019216.1 | XP_016874705.1 | 2 - 3 | 647 | 87 | ||
| GenBank mRNA | DB022290.1 | - | 2 - 3 | 196 | 87 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001256.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_006143.2,XM_011520623.2,XM_011520624.2,XM_011520625.2,XM_017019212.1,XM_017019213.1,XM_017019216.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200006888.hg.1 | ENST00000228872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006888.hg.1 | ENST00000228872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006888.hg.1 | ENST00000228872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006888.hg.1 | ENST00000228872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006888.hg.1 | ENST00000228872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006888.hg.1 | ENST00000228872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006888.hg.1 | ENST00000228872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006888.hg.1 | ENST00000228872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006888.hg.1 | ENST00000228872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006888.hg.1 | ENST00000228872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006888.hg.1 | ENST00000228872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006888.hg.1 | ENST00000228872.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006888.hg.1 | ENST00000396340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006888.hg.1 | ENST00000396340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006888.hg.1 | ENST00000396340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006888.hg.1 | ENST00000396340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006888.hg.1 | ENST00000396340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006888.hg.1 | ENST00000442489.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006888.hg.1 | ENST00000442489.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006888.hg.1 | ENST00000442489.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006888.hg.1 | ENST00000442489.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006888.hg.1 | ENST00000442489.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006888.hg.1 | ENST00000477087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006888.hg.1 | ENST00000477087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006888.hg.1 | ENST00000477087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006888.hg.1 | ENST00000477087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006888.hg.1 | ENST00000477087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006888.hg.1 | NM_004064.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006888.hg.1 | NM_004064.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006888.hg.1 | NM_004064.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006888.hg.1 | NM_004064.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006888.hg.1 | NM_004064.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006888.hg.1 | NM_004064.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006888.hg.1 | NM_004064.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006888.hg.1 | NM_004064.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006888.hg.1 | NM_004064.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006888.hg.1 | NM_004064.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006888.hg.1 | NM_004064.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009972.hg.1 | ENST00000332427.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009972.hg.1 | ENST00000332427.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009972.hg.1 | ENST00000332427.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009972.hg.1 | ENST00000332427.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009972.hg.1 | ENST00000332427.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009972.hg.1 | ENST00000332427.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009972.hg.1 | ENST00000332427.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009972.hg.1 | ENST00000332427.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009972.hg.1 | ENST00000537507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009972.hg.1 | ENST00000537507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009972.hg.1 | ENST00000537507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009972.hg.1 | ENST00000537507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009972.hg.1 | ENST00000540510.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009972.hg.1 | ENST00000540510.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009972.hg.1 | ENST00000540510.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009972.hg.1 | ENST00000540510.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009972.hg.1 | ENST00000540510.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009972.hg.1 | ENST00000540510.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009972.hg.1 | ENST00000540510.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009972.hg.1 | ENST00000540796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009972.hg.1 | ENST00000540796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009972.hg.1 | ENST00000540796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009972.hg.1 | ENST00000540796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009972.hg.1 | ENST00000540796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009972.hg.1 | ENST00000545876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009972.hg.1 | ENST00000545876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009972.hg.1 | ENST00000545876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009972.hg.1 | ENST00000545876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009972.hg.1 | ENST00000545876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009972.hg.1 | NM_006143.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009972.hg.1 | NM_006143.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009972.hg.1 | NM_006143.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009972.hg.1 | NM_006143.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009972.hg.1 | NM_006143.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009972.hg.1 | NM_006143.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009972.hg.1 | NM_006143.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009972.hg.1 | NM_006143.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12016420.hg.1 | ENST00000332427.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12016420.hg.1 | ENST00000537507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12016420.hg.1 | ENST00000540510.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12016420.hg.1 | NM_006143.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12016420.hg.1 | TC12001256.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1200006888.hg.1 | ENST00000228872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006888.hg.1 | ENST00000228872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006888.hg.1 | ENST00000228872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006888.hg.1 | ENST00000228872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006888.hg.1 | ENST00000228872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006888.hg.1 | ENST00000228872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006888.hg.1 | ENST00000228872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006888.hg.1 | ENST00000228872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006888.hg.1 | ENST00000228872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006888.hg.1 | ENST00000228872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006888.hg.1 | ENST00000228872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006888.hg.1 | ENST00000228872.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006888.hg.1 | ENST00000396340.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006888.hg.1 | ENST00000396340.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006888.hg.1 | ENST00000396340.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006888.hg.1 | ENST00000396340.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006888.hg.1 | ENST00000396340.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006888.hg.1 | ENST00000442489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006888.hg.1 | ENST00000442489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006888.hg.1 | ENST00000442489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006888.hg.1 | ENST00000442489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006888.hg.1 | ENST00000442489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006888.hg.1 | ENST00000477087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006888.hg.1 | ENST00000477087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006888.hg.1 | ENST00000477087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006888.hg.1 | ENST00000477087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006888.hg.1 | ENST00000477087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006888.hg.1 | NM_004064.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006888.hg.1 | NM_004064.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006888.hg.1 | NM_004064.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006888.hg.1 | NM_004064.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006888.hg.1 | NM_004064.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006888.hg.1 | NM_004064.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006888.hg.1 | NM_004064.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006888.hg.1 | NM_004064.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006888.hg.1 | NM_004064.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006888.hg.1 | NM_004064.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006888.hg.1 | NM_004064.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009972.hg.1 | ENST00000332427.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009972.hg.1 | ENST00000332427.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009972.hg.1 | ENST00000332427.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009972.hg.1 | ENST00000332427.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009972.hg.1 | ENST00000332427.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009972.hg.1 | ENST00000332427.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009972.hg.1 | ENST00000332427.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009972.hg.1 | ENST00000332427.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009972.hg.1 | ENST00000537507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009972.hg.1 | ENST00000537507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009972.hg.1 | ENST00000537507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009972.hg.1 | ENST00000537507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009972.hg.1 | ENST00000540510.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009972.hg.1 | ENST00000540510.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009972.hg.1 | ENST00000540510.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009972.hg.1 | ENST00000540510.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009972.hg.1 | ENST00000540510.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009972.hg.1 | ENST00000540510.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009972.hg.1 | ENST00000540510.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009972.hg.1 | ENST00000540796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009972.hg.1 | ENST00000540796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009972.hg.1 | ENST00000540796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009972.hg.1 | ENST00000540796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009972.hg.1 | ENST00000540796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009972.hg.1 | ENST00000545876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009972.hg.1 | ENST00000545876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009972.hg.1 | ENST00000545876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009972.hg.1 | ENST00000545876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009972.hg.1 | ENST00000545876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009972.hg.1 | NM_006143.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009972.hg.1 | NM_006143.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009972.hg.1 | NM_006143.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009972.hg.1 | NM_006143.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009972.hg.1 | NM_006143.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009972.hg.1 | NM_006143.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009972.hg.1 | NM_006143.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009972.hg.1 | NM_006143.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200175332.hg.1 | ENST00000332427.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200175332.hg.1 | ENST00000537507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200175332.hg.1 | ENST00000540510.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200175332.hg.1 | ENST00000540796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200175332.hg.1 | ENST00000545876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200175332.hg.1 | NM_006143.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200175332.hg.1 | ENST00000332427.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200175332.hg.1 | ENST00000537507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200175332.hg.1 | ENST00000540510.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200175332.hg.1 | ENST00000540796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200175332.hg.1 | ENST00000545876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200175332.hg.1 | NM_006143.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200175332.hg.1 | TC1200009972.hg.1 | Clariom D TC |

GPR19
Genomic Map