Assay Details
| Gene Symbol: MYHAS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_125367.1 | 7 - 8 | 640 | 136 | ||
| GenBank mRNA | AK097500.1 | - | 7 - 8 | 628 | 136 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | myosin heavy chain 2 | MYH2 |
IBM3, MYH2A, MYHSA2, MYHas8, MYPOP, MyHC-2A, MyHC-IIa |
-58.4kb | Anti-sense | Chr. 17 - 10521148 - 10549700 on NCBI Build GRCh38 |
| Downstream |
myosin, heavy chain 3, skeletal muscle, embryonic |
MYH3 |
DA2A, DA2B, DA8, HEMHC, MYHC-EMB, MYHSE1, SMHCE |
19.9kb | Anti-sense | Chr. 17 - 10628526 - 10679657 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000399342.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000399342.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000399342.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000399342.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000399342.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000399342.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000399342.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000399342.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000399342.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000399342.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000577181.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000577181.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000577181.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000577181.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000579914.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000579914.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000579914.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000581304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000581304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000581304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000581304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000581947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000581947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000581947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000581947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000581947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000581947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000581947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000581947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000581947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000581947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000581947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000581947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000585303.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000585303.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000585303.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000585303.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000585303.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000587182.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000587182.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000587182.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000587182.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000587182.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000587182.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000587182.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000587182.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000587182.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000587182.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000587182.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000587182.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000587182.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000587182.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | ENST00000587182.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | NR_125367.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | NR_125367.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | NR_125367.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | NR_125367.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | NR_125367.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | NR_125367.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | NR_125367.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | NR_125367.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | NR_125367.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | NR_125367.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | NR_125367.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | NR_125367.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | NR_125367.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | NR_125367.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | NR_125367.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | NR_125367.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | NR_125367.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | NR_125367.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | NR_125367.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006871.hg.1 | NR_125367.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148400.hg.1 | ENST00000579914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148400.hg.1 | ENST00000587182.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148400.hg.1 | NR_125367.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148400.hg.1 | TC1700006871.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700148400.hg.1 | ENST00000579914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148400.hg.1 | ENST00000587182.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148400.hg.1 | NR_125367.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000399342.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000399342.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000399342.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000399342.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000399342.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000399342.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000399342.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000399342.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000399342.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000399342.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000577181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000577181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000577181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000577181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000579914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000579914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000579914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000581304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000581304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000581304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000581304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000581947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000581947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000581947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000581947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000581947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000581947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000581947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000581947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000581947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000581947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000581947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000581947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000585303.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000585303.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000585303.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000585303.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000585303.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000587182.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000587182.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000587182.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000587182.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000587182.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000587182.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000587182.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000587182.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000587182.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000587182.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000587182.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000587182.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000587182.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000587182.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | ENST00000587182.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | NR_125367.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | NR_125367.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | NR_125367.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | NR_125367.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | NR_125367.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | NR_125367.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | NR_125367.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | NR_125367.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | NR_125367.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | NR_125367.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | NR_125367.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | NR_125367.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | NR_125367.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | NR_125367.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | NR_125367.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | NR_125367.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | NR_125367.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | NR_125367.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | NR_125367.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006871.hg.1 | NR_125367.1 | Refseq |

MYHAS
Genomic Map