Assay Details
| Gene Symbol: LOC101926948 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135130.1 | 1 - 2 | 161 | 82 | ||
| GenBank mRNA | BC042185.1 | - | 1 - 2 | 149 | 82 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | testis-specific kinase 1 | TESK1 | -36.2kb | Sense | Chr. 9 - 35605284 - 35610041 on NCBI Build GRCh38 | |
| Downstream | signaling threshold regulating transmembrane adaptor 1 | SIT1 |
SIT, SIT-R |
1.8kb | Anti-sense | Chr. 9 - 35649298 - 35650950 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC101926948
Genomic Map