Assay Details
| Gene Symbol: HS3ST3B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_130138.1 | 2 - 3 | 1702 | 79 | ||
| GenBank mRNA | AK023723.1 | - | 2 - 3 | 1669 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
glycosaminoglycan biosynthetic process
heparan sulfate proteoglycan biosynthetic process
heparan sulfate proteoglycan biosynthetic process, enzymatic modification
[heparan sulfate]-glucosamine 3-sulfotransferase 1 activity
[heparan sulfate]-glucosamine 3-sulfotransferase 3 activity
glycosaminoglycan biosynthetic process
heparan sulfate proteoglycan biosynthetic process
heparan sulfate proteoglycan biosynthetic process, enzymatic modification
[heparan sulfate]-glucosamine 3-sulfotransferase 1 activity
[heparan sulfate]-glucosamine 3-sulfotransferase 3 activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | ENST00000466596.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | ENST00000466596.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | ENST00000466596.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | ENST00000466596.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | ENST00000466596.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | ENST00000466596.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | ENST00000466596.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | ENST00000466596.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006946.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148940.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148940.hg.1 | ENST00000466596.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148940.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148940.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148940.hg.1 | TC1700006946.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700148940.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148940.hg.1 | ENST00000466596.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148940.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148940.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148944.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148944.hg.1 | ENST00000466596.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148944.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148944.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148944.hg.1 | TC1700006946.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700148944.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148944.hg.1 | ENST00000466596.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148944.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148944.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148945.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148945.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148945.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148945.hg.1 | TC1700006946.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700148945.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148945.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148945.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | ENST00000360954.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | ENST00000466596.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | ENST00000466596.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | ENST00000466596.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | ENST00000466596.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | ENST00000466596.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | ENST00000466596.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | ENST00000466596.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | ENST00000466596.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | NM_006041.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | NR_130138.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006946.hg.1 | NR_130138.1 | Refseq |

HS3ST3B1
Genomic Map