Assay Details
| Gene Symbol: LIPM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128215.1 | NP_001121687.1 | 3 - 4 | 632 | 81 | |
| XM_011539748.2 | XP_011538050.1 | 3 - 4 | 603 | 81 | ||
| XM_011539751.2 | XP_011538053.1 | 2 - 3 | 3972 | 81 | ||
| XM_011539753.2 | XP_011538055.1 | 3 - 4 | 603 | 81 | ||
| GenBank mRNA | BC157888.1 | - | 4 - 5 | 600 | 8862709 | 81 |
| BC171908.1 | - | 4 - 5 | 600 | 9054562 | 81 | |
| EF426484.1 | - | 3 - 4 | 622 | 81 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000630.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10006919.hg.1 | ENST00000404743.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006919.hg.1 | ENST00000539337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006919.hg.1 | NM_001128215.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006919.hg.1 | TC10000630.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006922.hg.1 | ENST00000404743.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006922.hg.1 | ENST00000539337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006922.hg.1 | NM_001128215.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006922.hg.1 | TC10000630.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006917.hg.1 | NM_001128215.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006917.hg.1 | TC10000630.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008383.hg.1 | ENST00000404743.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008383.hg.1 | ENST00000404743.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008383.hg.1 | ENST00000404743.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008383.hg.1 | ENST00000404743.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008383.hg.1 | ENST00000404743.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008383.hg.1 | ENST00000404743.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008383.hg.1 | ENST00000404743.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008383.hg.1 | ENST00000404743.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008383.hg.1 | ENST00000404743.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008383.hg.1 | ENST00000404743.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008383.hg.1 | ENST00000404743.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008383.hg.1 | ENST00000539337.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008383.hg.1 | ENST00000539337.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008383.hg.1 | ENST00000539337.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008383.hg.1 | ENST00000539337.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008383.hg.1 | ENST00000539337.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008383.hg.1 | ENST00000539337.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008383.hg.1 | ENST00000539337.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008383.hg.1 | ENST00000539337.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008383.hg.1 | ENST00000539337.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008383.hg.1 | ENST00000539337.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008383.hg.1 | NM_001128215.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008383.hg.1 | NM_001128215.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008383.hg.1 | NM_001128215.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008383.hg.1 | NM_001128215.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008383.hg.1 | NM_001128215.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008383.hg.1 | NM_001128215.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008383.hg.1 | NM_001128215.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008383.hg.1 | NM_001128215.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008383.hg.1 | NM_001128215.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008383.hg.1 | NM_001128215.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008383.hg.1 | NM_001128215.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008383.hg.1 | NM_001128215.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006918.hg.1 | ENST00000404743.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006918.hg.1 | NM_001128215.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006918.hg.1 | TC10000630.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006921.hg.1 | ENST00000404743.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006921.hg.1 | ENST00000539337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006921.hg.1 | NM_001128215.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006921.hg.1 | TC10000630.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006923.hg.1 | ENST00000404743.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006923.hg.1 | ENST00000539337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006923.hg.1 | NM_001128215.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006923.hg.1 | TC10000630.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1000156780.hg.1 | ENST00000404743.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156780.hg.1 | NM_001128215.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156780.hg.1 | ENST00000404743.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156780.hg.1 | NM_001128215.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156780.hg.1 | TC1000008383.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000156785.hg.1 | ENST00000404743.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156785.hg.1 | ENST00000539337.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156785.hg.1 | NM_001128215.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156785.hg.1 | ENST00000404743.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156785.hg.1 | ENST00000539337.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156785.hg.1 | NM_001128215.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156785.hg.1 | TC1000008383.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000008383.hg.1 | ENST00000404743.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008383.hg.1 | ENST00000404743.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008383.hg.1 | ENST00000404743.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008383.hg.1 | ENST00000404743.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008383.hg.1 | ENST00000404743.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008383.hg.1 | ENST00000404743.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008383.hg.1 | ENST00000404743.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008383.hg.1 | ENST00000404743.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008383.hg.1 | ENST00000404743.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008383.hg.1 | ENST00000404743.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008383.hg.1 | ENST00000404743.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008383.hg.1 | ENST00000539337.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008383.hg.1 | ENST00000539337.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008383.hg.1 | ENST00000539337.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008383.hg.1 | ENST00000539337.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008383.hg.1 | ENST00000539337.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008383.hg.1 | ENST00000539337.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008383.hg.1 | ENST00000539337.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008383.hg.1 | ENST00000539337.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008383.hg.1 | ENST00000539337.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008383.hg.1 | ENST00000539337.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008383.hg.1 | NM_001128215.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008383.hg.1 | NM_001128215.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008383.hg.1 | NM_001128215.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008383.hg.1 | NM_001128215.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008383.hg.1 | NM_001128215.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008383.hg.1 | NM_001128215.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008383.hg.1 | NM_001128215.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008383.hg.1 | NM_001128215.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008383.hg.1 | NM_001128215.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008383.hg.1 | NM_001128215.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008383.hg.1 | NM_001128215.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008383.hg.1 | NM_001128215.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156782.hg.1 | TC1000008383.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000156779.hg.1 | ENST00000404743.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156779.hg.1 | NM_001128215.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156779.hg.1 | ENST00000404743.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156779.hg.1 | NM_001128215.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156779.hg.1 | TC1000008383.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000156784.hg.1 | ENST00000404743.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156784.hg.1 | ENST00000539337.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156784.hg.1 | NM_001128215.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156784.hg.1 | ENST00000404743.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156784.hg.1 | ENST00000539337.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156784.hg.1 | NM_001128215.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156784.hg.1 | TC1000008383.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000156786.hg.1 | ENST00000404743.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156786.hg.1 | ENST00000539337.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156786.hg.1 | NM_001128215.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156786.hg.1 | ENST00000404743.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156786.hg.1 | ENST00000539337.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156786.hg.1 | NM_001128215.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156786.hg.1 | TC1000008383.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000156783.hg.1 | ENST00000539337.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156783.hg.1 | ENST00000539337.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156783.hg.1 | TC1000008383.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000156781.hg.1 | ENST00000404743.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156781.hg.1 | NM_001128215.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156781.hg.1 | ENST00000404743.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156781.hg.1 | NM_001128215.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156781.hg.1 | TC1000008383.hg.1 | Clariom D TC |

LIPM
Genomic Map