Assay Details
| Gene Symbol: YJEFN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198537.3 | NP_940939.2 | 1 - 2 | 131 | 70 | |
| GenBank mRNA | DQ778080.1 | - | 1 - 2 | 59 | 70 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000376.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19005077.hg.1 | ENST00000397179.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19005077.hg.1 | ENST00000436027.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19005077.hg.1 | NM_001190328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19005077.hg.1 | NM_198537.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19005077.hg.1 | TC19000376.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19005080.hg.1 | ENST00000397179.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19005080.hg.1 | ENST00000436027.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19005080.hg.1 | ENST00000510139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19005080.hg.1 | ENST00000514277.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19005080.hg.1 | NM_001190328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19005080.hg.1 | NM_198537.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19005080.hg.1 | TC19000376.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011689.hg.1 | ENST00000436027.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011689.hg.1 | ENST00000436027.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011689.hg.1 | ENST00000436027.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011689.hg.1 | ENST00000436027.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011689.hg.1 | ENST00000436027.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011689.hg.1 | ENST00000436027.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011689.hg.1 | ENST00000436027.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011689.hg.1 | ENST00000436027.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011689.hg.1 | ENST00000458210.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011689.hg.1 | ENST00000458210.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011689.hg.1 | ENST00000458210.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011689.hg.1 | ENST00000458210.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011689.hg.1 | ENST00000458210.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011689.hg.1 | ENST00000458210.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011689.hg.1 | ENST00000458210.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011689.hg.1 | ENST00000458210.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011689.hg.1 | ENST00000458210.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011689.hg.1 | ENST00000514277.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011689.hg.1 | ENST00000514277.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011689.hg.1 | ENST00000514277.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011689.hg.1 | ENST00000514277.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011689.hg.1 | ENST00000514277.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011689.hg.1 | ENST00000514277.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011689.hg.1 | ENST00000514277.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011689.hg.1 | ENST00000514277.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011689.hg.1 | ENST00000514277.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011689.hg.1 | NM_001190328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011689.hg.1 | NM_001190328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011689.hg.1 | NM_001190328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011689.hg.1 | NM_001190328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011689.hg.1 | NM_001190328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011689.hg.1 | NM_001190328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011689.hg.1 | NM_001190328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011689.hg.1 | NM_001190328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011689.hg.1 | NM_198537.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011689.hg.1 | NM_198537.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011689.hg.1 | NM_198537.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011689.hg.1 | NM_198537.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011689.hg.1 | NM_198537.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011689.hg.1 | NM_198537.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011689.hg.1 | NM_198537.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011689.hg.1 | NM_198537.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011689.hg.1 | NM_198537.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19005082.hg.1 | ENST00000397179.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19005082.hg.1 | ENST00000512771.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19005082.hg.1 | ENST00000514277.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19005082.hg.1 | NM_198537.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19005082.hg.1 | TC19000376.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1900011689.hg.1 | ENST00000436027.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011689.hg.1 | ENST00000436027.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011689.hg.1 | ENST00000436027.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011689.hg.1 | ENST00000436027.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011689.hg.1 | ENST00000436027.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011689.hg.1 | ENST00000436027.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011689.hg.1 | ENST00000436027.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011689.hg.1 | ENST00000436027.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011689.hg.1 | ENST00000458210.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011689.hg.1 | ENST00000458210.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011689.hg.1 | ENST00000458210.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011689.hg.1 | ENST00000458210.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011689.hg.1 | ENST00000458210.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011689.hg.1 | ENST00000458210.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011689.hg.1 | ENST00000458210.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011689.hg.1 | ENST00000458210.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011689.hg.1 | ENST00000458210.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011689.hg.1 | ENST00000514277.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011689.hg.1 | ENST00000514277.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011689.hg.1 | ENST00000514277.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011689.hg.1 | ENST00000514277.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011689.hg.1 | ENST00000514277.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011689.hg.1 | ENST00000514277.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011689.hg.1 | ENST00000514277.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011689.hg.1 | ENST00000514277.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011689.hg.1 | ENST00000514277.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011689.hg.1 | NM_001190328.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011689.hg.1 | NM_001190328.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011689.hg.1 | NM_001190328.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011689.hg.1 | NM_001190328.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011689.hg.1 | NM_001190328.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011689.hg.1 | NM_001190328.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011689.hg.1 | NM_001190328.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011689.hg.1 | NM_001190328.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011689.hg.1 | NM_198537.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011689.hg.1 | NM_198537.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011689.hg.1 | NM_198537.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011689.hg.1 | NM_198537.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011689.hg.1 | NM_198537.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011689.hg.1 | NM_198537.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011689.hg.1 | NM_198537.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011689.hg.1 | NM_198537.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011689.hg.1 | NM_198537.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900199934.hg.1 | ENST00000514277.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199934.hg.1 | NM_198537.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900199934.hg.1 | TC1900011689.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900199934.hg.1 | ENST00000514277.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199934.hg.1 | NM_198537.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900199932.hg.1 | ENST00000436027.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199932.hg.1 | ENST00000458210.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199932.hg.1 | ENST00000514277.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199932.hg.1 | NM_001190328.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900199932.hg.1 | NM_198537.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900199932.hg.1 | TC1900011689.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900199932.hg.1 | ENST00000436027.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199932.hg.1 | ENST00000458210.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199932.hg.1 | ENST00000514277.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199932.hg.1 | NM_001190328.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900199932.hg.1 | NM_198537.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900199933.hg.1 | ENST00000436027.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199933.hg.1 | ENST00000458210.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199933.hg.1 | ENST00000514277.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199933.hg.1 | NM_001190328.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900199933.hg.1 | NM_198537.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900199933.hg.1 | TC1900011689.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900199933.hg.1 | ENST00000436027.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199933.hg.1 | ENST00000458210.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199933.hg.1 | ENST00000514277.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900199933.hg.1 | NM_001190328.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900199933.hg.1 | NM_198537.3 | Refseq |

YJEFN3
Genomic Map