Assay Details
| Gene Symbol: YJEFN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190328.1 | NP_001177257.1 | 5 - 6 | 621 | 65 | |
| NM_198537.3 | NP_940939.2 | 6 - 7 | 771 | 65 | ||
| XM_005259903.4 | XP_005259960.1 | 5 - 6 | 1103 | 65 | ||
| XM_011527997.1 | XP_011526299.1 | 6 - 7 | 1037 | 65 | ||
| XM_011527998.2 | XP_011526300.1 | 6 - 7 | 1317 | 65 | ||
| XM_011527999.1 | XP_011526301.1 | 6 - 7 | 846 | 65 | ||
| XM_017026770.1 | XP_016882259.1 | 5 - 6 | 819 | 65 | ||
| XM_017026771.1 | XP_016882260.1 | 5 - 6 | 1400 | 65 | ||
| GenBank mRNA | AK094976.1 | - | 3 - 4 | 1734 | 65 | |
| AK126916.1 | - | 5 - 6 | 2510 | 65 | ||
| BI520231.1 | - | 5 - 6 | 622 | 65 | ||
| BX097402.1 | - | 1 - 2 | 276 | 65 | ||
| DB551327.1 | - | 2 - 3 | 224 | 65 | ||
| DQ778079.1 | - | 5 - 6 | 549 | 65 | ||
| DQ778080.1 | - | 6 - 7 | 699 | 65 | ||
More Information
Set Membership: |
|

YJEFN3
Genomic Map