Assay Details
| Gene Symbol: ZNF213-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC013599.1 | - | 1 - 2 | 293 | 63 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000570901.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000570901.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000570901.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000570901.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000573414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000573414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000573414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000573447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000573447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000573447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000573447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000573447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000573447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000573447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000573447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000573447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000573447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000573447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000575089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000575089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000575089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000575089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000575089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000576590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000576590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000576590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000576590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000577163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000577163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000577163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000577163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000577163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | NR_110900.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | NR_110900.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | NR_110900.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | NR_110900.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | NR_110900.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | NR_110900.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | NR_110900.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | NR_110900.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | NR_110900.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | NR_110900.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000570901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000570901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000570901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000570901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000573414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000573414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000573414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000573447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000573447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000573447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000573447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000573447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000573447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000573447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000573447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000573447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000573447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000573447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000575089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000575089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000575089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000575089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000575089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000576590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000576590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000576590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000576590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000577163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000577163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000577163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000577163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000577163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | NR_110900.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | NR_110900.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | NR_110900.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | NR_110900.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | NR_110900.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | NR_110900.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | NR_110900.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | NR_110900.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | NR_110900.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | NR_110900.1 | Refseq |

ZNF213-AS1
Genomic Map