Assay Details
| Gene Symbol: ADRA1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BX648936.1 | - | 1 | 547 | 97 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101927790 | LOC101927790 | -12.7kb | Anti-sense | Chr. 5 - 159936619 - 159941617 on NCBI Build GRCh38 | |
| Downstream | tetratricopeptide repeat domain 1 | TTC1 | TPR1 | 58.2kb | Sense | Chr. 5 - 160009100 - 160065545 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

ADRA1B
Genomic Map