| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01045288.hg.1 |
ENST00000287474.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01045288.hg.1 |
ENST00000414213.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01045288.hg.1 |
NM_001013660.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01045288.hg.1 |
TC01002911.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015049.hg.1 |
ENST00000287474.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015049.hg.1 |
ENST00000287474.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015049.hg.1 |
ENST00000287474.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015049.hg.1 |
ENST00000287474.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015049.hg.1 |
ENST00000287474.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015049.hg.1 |
ENST00000287474.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015049.hg.1 |
ENST00000287474.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015049.hg.1 |
ENST00000287474.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015049.hg.1 |
ENST00000287474.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015049.hg.1 |
ENST00000287474.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015049.hg.1 |
ENST00000287474.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015049.hg.1 |
ENST00000287474.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015049.hg.1 |
ENST00000287474.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015049.hg.1 |
ENST00000287474.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015049.hg.1 |
ENST00000287474.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015049.hg.1 |
ENST00000287474.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015049.hg.1 |
ENST00000287474.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015049.hg.1 |
ENST00000287474.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015049.hg.1 |
ENST00000287474.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015049.hg.1 |
ENST00000370176.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015049.hg.1 |
ENST00000370176.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015049.hg.1 |
ENST00000370176.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015049.hg.1 |
ENST00000370176.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015049.hg.1 |
ENST00000489209.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015049.hg.1 |
ENST00000489209.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015049.hg.1 |
ENST00000489209.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015049.hg.1 |
ENST00000492943.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015049.hg.1 |
ENST00000492943.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015049.hg.1 |
ENST00000492943.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015049.hg.1 |
ENST00000492943.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015049.hg.1 |
ENST00000492943.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015049.hg.1 |
NM_001013660.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015049.hg.1 |
NM_001013660.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015049.hg.1 |
NM_001013660.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015049.hg.1 |
NM_001013660.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015049.hg.1 |
NM_001013660.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015049.hg.1 |
NM_001013660.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015049.hg.1 |
NM_001013660.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015049.hg.1 |
NM_001013660.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015049.hg.1 |
NM_001013660.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015049.hg.1 |
NM_001013660.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015049.hg.1 |
NM_001013660.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015049.hg.1 |
NM_001013660.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015049.hg.1 |
NM_001013660.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015049.hg.1 |
NM_001013660.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015049.hg.1 |
NM_001013660.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015049.hg.1 |
NM_001013660.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015049.hg.1 |
NM_001013660.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015049.hg.1 |
NM_001013660.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015049.hg.1 |
NM_001013660.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100212131.hg.1 |
ENST00000287474.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100212131.hg.1 |
NM_001013660.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100212131.hg.1 |
ENST00000287474.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100212131.hg.1 |
NM_001013660.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100212131.hg.1 |
TC0100015049.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100015049.hg.1 |
ENST00000287474.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015049.hg.1 |
ENST00000287474.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015049.hg.1 |
ENST00000287474.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015049.hg.1 |
ENST00000287474.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015049.hg.1 |
ENST00000287474.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015049.hg.1 |
ENST00000287474.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015049.hg.1 |
ENST00000287474.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015049.hg.1 |
ENST00000287474.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015049.hg.1 |
ENST00000287474.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015049.hg.1 |
ENST00000287474.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015049.hg.1 |
ENST00000287474.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015049.hg.1 |
ENST00000287474.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015049.hg.1 |
ENST00000287474.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015049.hg.1 |
ENST00000287474.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015049.hg.1 |
ENST00000287474.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015049.hg.1 |
ENST00000287474.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015049.hg.1 |
ENST00000287474.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015049.hg.1 |
ENST00000287474.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015049.hg.1 |
ENST00000287474.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015049.hg.1 |
ENST00000370176.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015049.hg.1 |
ENST00000370176.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015049.hg.1 |
ENST00000370176.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015049.hg.1 |
ENST00000370176.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015049.hg.1 |
ENST00000489209.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015049.hg.1 |
ENST00000489209.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015049.hg.1 |
ENST00000489209.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015049.hg.1 |
ENST00000492943.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015049.hg.1 |
ENST00000492943.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015049.hg.1 |
ENST00000492943.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015049.hg.1 |
ENST00000492943.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015049.hg.1 |
ENST00000492943.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015049.hg.1 |
NM_001013660.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015049.hg.1 |
NM_001013660.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015049.hg.1 |
NM_001013660.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015049.hg.1 |
NM_001013660.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015049.hg.1 |
NM_001013660.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015049.hg.1 |
NM_001013660.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015049.hg.1 |
NM_001013660.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015049.hg.1 |
NM_001013660.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015049.hg.1 |
NM_001013660.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015049.hg.1 |
NM_001013660.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015049.hg.1 |
NM_001013660.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015049.hg.1 |
NM_001013660.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015049.hg.1 |
NM_001013660.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015049.hg.1 |
NM_001013660.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015049.hg.1 |
NM_001013660.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015049.hg.1 |
NM_001013660.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015049.hg.1 |
NM_001013660.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015049.hg.1 |
NM_001013660.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015049.hg.1 |
NM_001013660.2 |
Refseq |