Assay Details
| Gene Symbol: FRRS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013660.2 | NP_001013682.2 | 10 - 11 | 1728 | 88 | |
| XM_005270861.3 | XP_005270918.1 | 10 - 11 | 1255 | 88 | ||
| XM_011541451.2 | XP_011539753.1 | 10 - 11 | 1728 | 88 | ||
| XM_011541452.1 | XP_011539754.1 | 10 - 11 | 1255 | 88 | ||
| XM_017001271.1 | XP_016856760.1 | 8 - 9 | 1139 | 88 | ||
| GenBank mRNA | AK131302.1 | - | 10 - 11 | 1728 | 88 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002911.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01045281.hg.1 | ENST00000287474.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045281.hg.1 | ENST00000414213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045281.hg.1 | NM_001013660.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045281.hg.1 | TC01002911.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045282.hg.1 | ENST00000287474.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045282.hg.1 | ENST00000414213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045282.hg.1 | NM_001013660.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045282.hg.1 | TC01002911.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000370176.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000370176.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000370176.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000370176.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000489209.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000489209.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000489209.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000492943.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000492943.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000492943.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000492943.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000492943.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045283.hg.1 | ENST00000287474.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045283.hg.1 | ENST00000414213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045283.hg.1 | NM_001013660.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045283.hg.1 | TC01002911.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0100212126.hg.1 | ENST00000287474.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100212126.hg.1 | NM_001013660.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100212126.hg.1 | ENST00000287474.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100212126.hg.1 | NM_001013660.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100212126.hg.1 | TC0100015049.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000370176.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000370176.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000370176.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000370176.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000489209.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000489209.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000489209.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000492943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000492943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000492943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000492943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000492943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100212125.hg.1 | ENST00000287474.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100212125.hg.1 | NM_001013660.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100212125.hg.1 | ENST00000287474.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100212125.hg.1 | NM_001013660.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100212125.hg.1 | TC0100015049.hg.1 | Clariom D TC |

FRRS1
Genomic Map