Assay Details
| Gene Symbol: KBTBD12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207335.2 | NP_997218.2 | 2 - 3 | 1345 | 115 | |
| XM_005247167.1 | XP_005247224.1 | 3 - 4 | 1809 | 115 | ||
| XM_005247168.4 | XP_005247225.1 | 3 - 4 | 1887 | 115 | ||
| XM_011512490.2 | XP_011510792.1 | 3 - 4 | 1809 | 115 | ||
| XM_011512491.2 | XP_011510793.1 | 3 - 4 | 1809 | 115 | ||
| XM_011512492.2 | XP_011510794.1 | 3 - 4 | 1809 | 115 | ||
| GenBank mRNA | BX537865.1 | - | 2 - 3 | 384 | 115 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000668.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03012045.hg.1 | ENST00000343941.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03012045.hg.1 | ENST00000405109.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03012045.hg.1 | ENST00000405256.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03012045.hg.1 | ENST00000407609.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03012045.hg.1 | ENST00000492025.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03012045.hg.1 | NM_207335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03012045.hg.1 | TC03000668.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000343941.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000343941.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000343941.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000343941.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000343941.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000343941.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000343941.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000405256.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000405256.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000405256.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000405256.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000405256.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000405256.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000405256.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000405256.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000405256.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000405256.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000405256.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000407609.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000407609.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000407609.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000407609.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000407609.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000407609.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000407609.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000407609.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000476626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000476626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000476626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000476626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000492025.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000492025.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000492025.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000492025.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000497045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000497045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000497045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000497045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000497045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000497045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | ENST00000497045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | NM_207335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | NM_207335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | NM_207335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | NM_207335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | NM_207335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | NM_207335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | NM_207335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | NM_207335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | NM_207335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | NM_207335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008711.hg.1 | NM_207335.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000343941.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000343941.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000343941.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000343941.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000343941.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000343941.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000343941.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000405256.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000405256.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000405256.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000405256.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000405256.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000405256.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000405256.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000405256.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000405256.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000405256.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000405256.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000407609.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000407609.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000407609.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000407609.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000407609.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000407609.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000407609.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000407609.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000476626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000476626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000476626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000476626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000492025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000492025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000492025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000492025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000497045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000497045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000497045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000497045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000497045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000497045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | ENST00000497045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | NM_207335.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | NM_207335.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | NM_207335.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | NM_207335.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | NM_207335.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | NM_207335.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | NM_207335.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | NM_207335.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | NM_207335.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | NM_207335.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008711.hg.1 | NM_207335.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300161932.hg.1 | ENST00000343941.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161932.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161932.hg.1 | ENST00000405256.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161932.hg.1 | ENST00000407609.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161932.hg.1 | ENST00000492025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161932.hg.1 | NM_207335.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300161932.hg.1 | ENST00000343941.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161932.hg.1 | ENST00000405109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161932.hg.1 | ENST00000405256.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161932.hg.1 | ENST00000407609.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161932.hg.1 | ENST00000492025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161932.hg.1 | NM_207335.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300161932.hg.1 | TC0300008711.hg.1 | Clariom D TC |

KBTBD12
Genomic Map