Assay Details
| Gene Symbol: LOC101927021 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110070.1 | 3 - 4 | 359 | 185 | ||
| GenBank mRNA | BM544796.1 | - | 3 - 4 | 380 | 185 | |
More Information
Updated Mapping Information
Set Membership: |
|

LOC101927021
Genomic Map