Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101927854 | LOC101927854 | -265.2kb | Anti-sense | Chr. 3 - 24151324 - 24189956 on NCBI Build GRCh38 | |
| Downstream | THRB antisense RNA 1 | THRB-AS1 | 34.7kb | Anti-sense | Chr. 3 - 24494087 - 24500011 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map