| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010248.hg.1 |
ENST00000412578.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010248.hg.1 |
ENST00000412578.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010248.hg.1 |
ENST00000412578.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010248.hg.1 |
ENST00000412578.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010248.hg.1 |
ENST00000412578.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010248.hg.1 |
ENST00000412578.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010248.hg.1 |
ENST00000412578.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010248.hg.1 |
ENST00000436060.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010248.hg.1 |
ENST00000436060.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010248.hg.1 |
ENST00000436060.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010248.hg.1 |
ENST00000436060.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010248.hg.1 |
ENST00000436060.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010248.hg.1 |
NR_027102.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010248.hg.1 |
NR_027102.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010248.hg.1 |
NR_027102.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010248.hg.1 |
NR_027102.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010248.hg.1 |
NR_027102.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010248.hg.1 |
NR_027102.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010248.hg.1 |
NR_027102.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03020141.hg.1 |
ENST00000412578.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03020141.hg.1 |
ENST00000436060.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03020141.hg.1 |
NR_027102.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03020141.hg.1 |
TC03001164.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03020140.hg.1 |
ENST00000412578.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03020140.hg.1 |
NR_027102.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03020140.hg.1 |
TC03001164.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0300174844.hg.1 |
ENST00000412578.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300174844.hg.1 |
NR_027102.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300174844.hg.1 |
ENST00000412578.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300174844.hg.1 |
NR_027102.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300174844.hg.1 |
TC0300010248.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300174848.hg.1 |
ENST00000412578.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300174848.hg.1 |
ENST00000436060.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300174848.hg.1 |
NR_027102.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300174848.hg.1 |
ENST00000412578.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300174848.hg.1 |
ENST00000436060.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300174848.hg.1 |
NR_027102.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300174848.hg.1 |
TC0300010248.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300174847.hg.1 |
ENST00000412578.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300174847.hg.1 |
ENST00000436060.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300174847.hg.1 |
NR_027102.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300174847.hg.1 |
ENST00000412578.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300174847.hg.1 |
ENST00000436060.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300174847.hg.1 |
NR_027102.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300174847.hg.1 |
TC0300010248.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0300010248.hg.1 |
ENST00000412578.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010248.hg.1 |
ENST00000412578.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010248.hg.1 |
ENST00000412578.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010248.hg.1 |
ENST00000412578.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010248.hg.1 |
ENST00000412578.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010248.hg.1 |
ENST00000412578.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010248.hg.1 |
ENST00000412578.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010248.hg.1 |
ENST00000436060.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010248.hg.1 |
ENST00000436060.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010248.hg.1 |
ENST00000436060.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010248.hg.1 |
ENST00000436060.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010248.hg.1 |
ENST00000436060.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010248.hg.1 |
NR_027102.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010248.hg.1 |
NR_027102.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010248.hg.1 |
NR_027102.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010248.hg.1 |
NR_027102.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010248.hg.1 |
NR_027102.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010248.hg.1 |
NR_027102.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010248.hg.1 |
NR_027102.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300174846.hg.1 |
ENST00000412578.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300174846.hg.1 |
ENST00000436060.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300174846.hg.1 |
NR_027102.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300174846.hg.1 |
ENST00000412578.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300174846.hg.1 |
ENST00000436060.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300174846.hg.1 |
NR_027102.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300174846.hg.1 |
TC0300010248.hg.1 |
Clariom D TC |