| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07026671.hg.1 |
ENST00000306312.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07026671.hg.1 |
ENST00000339444.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07026671.hg.1 |
ENST00000354356.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07026671.hg.1 |
ENST00000356767.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07026671.hg.1 |
ENST00000393723.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07026671.hg.1 |
ENST00000393727.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07026671.hg.1 |
ENST00000393729.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07026671.hg.1 |
ENST00000393730.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07026671.hg.1 |
ENST00000393735.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07026671.hg.1 |
ENST00000432958.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07026671.hg.1 |
NM_001167962.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07026671.hg.1 |
NM_198999.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07026671.hg.1 |
NM_206883.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07026671.hg.1 |
NM_206884.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07026671.hg.1 |
NM_206885.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07026671.hg.1 |
TC07001719.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0700187300.hg.1 |
ENST00000306312.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700187300.hg.1 |
ENST00000339444.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700187300.hg.1 |
ENST00000354356.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700187300.hg.1 |
ENST00000356767.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700187300.hg.1 |
ENST00000393723.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700187300.hg.1 |
ENST00000393727.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700187300.hg.1 |
ENST00000393729.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700187300.hg.1 |
ENST00000393730.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700187300.hg.1 |
ENST00000393735.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700187300.hg.1 |
ENST00000423416.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700187300.hg.1 |
ENST00000432958.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700187300.hg.1 |
ENST00000445809.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700187300.hg.1 |
ENST00000454864.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700187300.hg.1 |
ENST00000456463.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700187300.hg.1 |
ENST00000487407.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700187300.hg.1 |
NM_001167962.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700187300.hg.1 |
NM_198999.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700187300.hg.1 |
NM_206883.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700187300.hg.1 |
NM_206884.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700187300.hg.1 |
NM_206885.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700187300.hg.1 |
NR_120441.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700187300.hg.1 |
NR_120442.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700187300.hg.1 |
NR_120443.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700187300.hg.1 |
ENST00000306312.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700187300.hg.1 |
ENST00000339444.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700187300.hg.1 |
ENST00000354356.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700187300.hg.1 |
ENST00000356767.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700187300.hg.1 |
ENST00000393723.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700187300.hg.1 |
ENST00000393727.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700187300.hg.1 |
ENST00000393729.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700187300.hg.1 |
ENST00000393730.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700187300.hg.1 |
ENST00000393735.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700187300.hg.1 |
ENST00000423416.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700187300.hg.1 |
ENST00000432958.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700187300.hg.1 |
ENST00000445809.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700187300.hg.1 |
ENST00000454864.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700187300.hg.1 |
ENST00000456463.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700187300.hg.1 |
ENST00000487407.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700187300.hg.1 |
NM_001167962.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700187300.hg.1 |
NM_198999.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700187300.hg.1 |
NM_206883.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700187300.hg.1 |
NM_206884.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700187300.hg.1 |
NM_206885.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700187300.hg.1 |
NR_120441.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700187300.hg.1 |
NR_120442.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700187300.hg.1 |
NR_120443.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700187300.hg.1 |
TC0700012123.hg.1 |
Clariom D TC |