Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC100507053 | LOC100507053 | Anti-sense | Chr. 4 - 99088857 - 99301356 on NCBI Build GRCh38 | ||
| Downstream |
alcohol dehydrogenase 4 (class II), pi polypeptide |
ADH4 |
ADH-2, HEL-S-4 |
21.0kb | Sense | Chr. 4 - 99123657 - 99144330 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map