| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10012341.hg.1 |
NR_026743.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10012341.hg.1 |
TC10000983.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10012342.hg.1 |
NR_026743.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10012342.hg.1 |
TC10000983.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009647.hg.1 |
ENST00000432950.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009647.hg.1 |
ENST00000432950.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009647.hg.1 |
ENST00000432950.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009647.hg.1 |
ENST00000432950.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009647.hg.1 |
ENST00000432950.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009647.hg.1 |
ENST00000432950.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009647.hg.1 |
ENST00000432950.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009647.hg.1 |
ENST00000432950.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009647.hg.1 |
ENST00000432950.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009647.hg.1 |
NR_026743.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009647.hg.1 |
NR_026743.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009647.hg.1 |
NR_026743.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009647.hg.1 |
NR_026743.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009647.hg.1 |
NR_026743.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009647.hg.1 |
NR_026743.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009647.hg.1 |
NR_026743.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009647.hg.1 |
NR_026743.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009647.hg.1 |
NR_026743.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009647.hg.1 |
NR_026743.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009647.hg.1 |
NR_026743.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009647.hg.1 |
NR_026743.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009647.hg.1 |
NR_026743.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009663.hg.1 |
ENST00000305623.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009663.hg.1 |
ENST00000305623.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009663.hg.1 |
ENST00000305623.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009663.hg.1 |
ENST00000305623.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009663.hg.1 |
ENST00000305623.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009663.hg.1 |
ENST00000305623.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009663.hg.1 |
ENST00000305623.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009663.hg.1 |
ENST00000305623.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009663.hg.1 |
ENST00000305623.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009663.hg.1 |
ENST00000305623.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009663.hg.1 |
ENST00000305623.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009663.hg.1 |
ENST00000305623.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009663.hg.1 |
ENST00000432689.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009663.hg.1 |
ENST00000432689.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009663.hg.1 |
ENST00000432689.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009663.hg.1 |
ENST00000432689.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009663.hg.1 |
ENST00000432689.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009663.hg.1 |
ENST00000432689.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009663.hg.1 |
ENST00000432689.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009663.hg.1 |
ENST00000432689.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000165901.hg.1 |
ENST00000432950.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000165901.hg.1 |
NR_026743.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000165901.hg.1 |
ENST00000432950.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000165901.hg.1 |
NR_026743.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000165901.hg.1 |
TC1000009647.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000165899.hg.1 |
ENST00000432950.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000165899.hg.1 |
NR_026743.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000165899.hg.1 |
ENST00000432950.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000165899.hg.1 |
NR_026743.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000165899.hg.1 |
TC1000009647.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1000009663.hg.1 |
ENST00000305623.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009663.hg.1 |
ENST00000305623.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009663.hg.1 |
ENST00000305623.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009663.hg.1 |
ENST00000305623.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009663.hg.1 |
ENST00000305623.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009663.hg.1 |
ENST00000305623.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009663.hg.1 |
ENST00000305623.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009663.hg.1 |
ENST00000305623.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009663.hg.1 |
ENST00000305623.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009663.hg.1 |
ENST00000305623.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009663.hg.1 |
ENST00000305623.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009663.hg.1 |
ENST00000305623.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009663.hg.1 |
ENST00000432689.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009663.hg.1 |
ENST00000432689.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009663.hg.1 |
ENST00000432689.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009663.hg.1 |
ENST00000432689.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009663.hg.1 |
ENST00000432689.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009663.hg.1 |
ENST00000432689.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009663.hg.1 |
ENST00000432689.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009663.hg.1 |
ENST00000432689.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009647.hg.1 |
ENST00000432950.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009647.hg.1 |
ENST00000432950.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009647.hg.1 |
ENST00000432950.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009647.hg.1 |
ENST00000432950.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009647.hg.1 |
ENST00000432950.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009647.hg.1 |
ENST00000432950.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009647.hg.1 |
ENST00000432950.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009647.hg.1 |
ENST00000432950.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009647.hg.1 |
ENST00000432950.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009647.hg.1 |
NR_026743.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009647.hg.1 |
NR_026743.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009647.hg.1 |
NR_026743.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009647.hg.1 |
NR_026743.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009647.hg.1 |
NR_026743.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009647.hg.1 |
NR_026743.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009647.hg.1 |
NR_026743.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009647.hg.1 |
NR_026743.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009647.hg.1 |
NR_026743.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009647.hg.1 |
NR_026743.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009647.hg.1 |
NR_026743.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009647.hg.1 |
NR_026743.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009647.hg.1 |
NR_026743.1 |
Refseq |