Assay Details
| Gene Symbol: MAP3K10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002446.3 | NP_002437.2 | 3 - 4 | 1304 | 86 | |
| GenBank mRNA | X90846.1 | - | 3 - 4 | 1304 | 86 | |
| Z48615.1 | - | 3 - 4 | 1176 | 86 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000559.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
signal transduction
smoothened signaling pathway
JNK cascade
activation of JNKK activity
activation of JUN kinase activity
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
positive regulation of apoptotic process
negative regulation of sequence-specific DNA binding transcription factor activity
positive regulation of JUN kinase activity
negative regulation of transcription, DNA-templated
positive regulation of JNK cascade
protein autophosphorylation
transcription corepressor activity
protein kinase activity
protein serine/threonine kinase activity
JUN kinase kinase kinase activity
ATP binding
protein homodimerization activity
bHLH transcription factor binding
apoptotic process
signal transduction
smoothened signaling pathway
JNK cascade
activation of JNKK activity
activation of JUN kinase activity
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
positive regulation of apoptotic process
negative regulation of sequence-specific DNA binding transcription factor activity
positive regulation of JUN kinase activity
negative regulation of transcription, DNA-templated
positive regulation of JNK cascade
protein autophosphorylation
transcription corepressor activity
protein kinase activity
protein serine/threonine kinase activity
JUN kinase kinase kinase activity
ATP binding
protein homodimerization activity
bHLH transcription factor binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000593502.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000593502.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000593502.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000593502.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000593502.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000593502.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000593502.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000593906.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000593906.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000593906.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000593906.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000593906.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000594569.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000594569.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000594569.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000594569.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000594791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000594791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000594791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000594791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000594791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000594791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000594951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000594951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000594951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000594951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000594951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000594951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000594951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000594951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000597986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000597986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000597986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000597986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000597986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000597986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000597986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000597986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000597986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000597986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000597986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000597986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000601702.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000601702.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | ENST00000601702.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | NM_002446.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | NM_002446.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | NM_002446.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | NM_002446.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | NM_002446.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | NM_002446.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | NM_002446.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | NM_002446.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | NM_002446.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | NM_002446.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | NM_002446.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | NM_002446.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | NM_002446.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | NM_002446.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | NM_002446.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | NM_002446.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | NM_002446.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | NM_002446.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | NM_002446.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | NM_002446.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008090.hg.1 | NM_002446.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19007613.hg.1 | ENST00000253055.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19007613.hg.1 | NM_002446.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19007613.hg.1 | TC19000559.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000253055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000593502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000593502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000593502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000593502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000593502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000593502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000593502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000593906.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000593906.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000593906.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000593906.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000593906.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000594569.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000594569.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000594569.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000594569.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000594791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000594791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000594791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000594791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000594791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000594791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000594951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000594951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000594951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000594951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000594951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000594951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000594951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000594951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000597986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000597986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000597986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000597986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000597986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000597986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000597986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000597986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000597986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000597986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000597986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000597986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000601702.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000601702.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | ENST00000601702.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | NM_002446.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | NM_002446.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | NM_002446.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | NM_002446.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | NM_002446.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | NM_002446.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | NM_002446.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | NM_002446.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | NM_002446.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | NM_002446.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | NM_002446.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | NM_002446.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | NM_002446.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | NM_002446.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | NM_002446.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | NM_002446.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | NM_002446.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | NM_002446.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | NM_002446.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | NM_002446.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008090.hg.1 | NM_002446.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900161047.hg.1 | ENST00000253055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900161047.hg.1 | ENST00000593502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900161047.hg.1 | ENST00000594951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900161047.hg.1 | ENST00000597986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900161047.hg.1 | ENST00000601702.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900161047.hg.1 | NM_002446.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900161047.hg.1 | TC1900008090.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900161047.hg.1 | ENST00000253055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900161047.hg.1 | ENST00000593502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900161047.hg.1 | ENST00000594951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900161047.hg.1 | ENST00000597986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900161047.hg.1 | ENST00000601702.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900161047.hg.1 | NM_002446.3 | Refseq |

MAP3K10
Genomic Map