Assay Details
| Gene Symbol: HSD17B6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011538925.1 | XP_011537227.1 | 1 - 2 | 135 | 72 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011538925.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
androgen biosynthetic process
androgen catabolic process
steroid metabolic process
oxidation-reduction process
catalytic activity
estradiol 17-beta-dehydrogenase activity
retinol dehydrogenase activity
electron carrier activity
oxidoreductase activity
testosterone dehydrogenase (NAD+) activity
androgen biosynthetic process
androgen catabolic process
steroid metabolic process
oxidation-reduction process
catalytic activity
estradiol 17-beta-dehydrogenase activity
retinol dehydrogenase activity
electron carrier activity
oxidoreductase activity
testosterone dehydrogenase (NAD+) activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000322165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000322165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000322165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000322165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000322165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000322165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000322165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000322165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000322165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000322165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000322165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000322165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000553476.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000553476.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000554150.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000554150.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000554150.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000554150.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000554150.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000554150.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000554150.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000554150.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000554150.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000554150.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000554150.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000554150.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000554155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000554155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000554155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000554155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000554155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000554643.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000554643.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000554643.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000554643.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000554643.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000554643.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000554643.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000554643.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000554643.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000554643.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000554643.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000554643.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000554643.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000554643.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000554643.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000555159.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000555159.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000555159.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000555159.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000555159.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000555159.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000555159.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000555159.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000555159.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000555159.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000555159.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000555805.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000555805.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000555805.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000555805.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000555805.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000555805.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000555805.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000555805.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000555805.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000555805.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000555805.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000555805.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000555805.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000556481.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000556481.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000556481.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000556481.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000556481.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000556650.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000556650.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000556650.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000556650.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000556650.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000556875.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000556875.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000556875.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | ENST00000556875.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | NM_003725.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | NM_003725.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | NM_003725.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | NM_003725.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | NM_003725.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | NM_003725.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | NM_003725.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | NM_003725.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | NM_003725.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | NM_003725.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | NM_003725.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | NM_003725.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007847.hg.1 | NM_003725.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000322165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000322165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000322165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000322165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000322165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000322165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000322165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000322165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000322165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000322165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000322165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000322165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000553476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000553476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000554150.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000554150.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000554150.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000554150.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000554150.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000554150.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000554150.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000554150.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000554150.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000554150.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000554150.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000554150.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000554155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000554155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000554155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000554155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000554155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000554643.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000554643.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000554643.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000554643.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000554643.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000554643.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000554643.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000554643.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000554643.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000554643.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000554643.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000554643.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000554643.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000554643.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000554643.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000555159.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000555159.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000555159.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000555159.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000555159.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000555159.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000555159.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000555159.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000555159.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000555159.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000555159.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000555805.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000555805.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000555805.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000555805.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000555805.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000555805.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000555805.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000555805.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000555805.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000555805.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000555805.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000555805.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000555805.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000556481.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000556481.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000556481.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000556481.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000556481.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000556650.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000556650.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000556650.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000556650.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000556650.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000556875.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000556875.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000556875.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | ENST00000556875.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | NM_003725.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | NM_003725.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | NM_003725.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | NM_003725.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | NM_003725.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | NM_003725.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | NM_003725.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | NM_003725.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | NM_003725.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | NM_003725.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | NM_003725.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | NM_003725.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007847.hg.1 | NM_003725.3 | Refseq |

HSD17B6
Genomic Map