Assay Details
| Gene Symbol: ATP6V1C2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039362.1 | NP_001034451.1 | 12 - 13 | 1166 | 61 | |
| NM_144583.3 | NP_653184.2 | 11 - 12 | 1028 | 61 | ||
| XM_011510339.2 | XP_011508641.1 | 13 - 14 | 1163 | 61 | ||
| XM_011510340.2 | XP_011508642.1 | 12 - 13 | 1025 | 61 | ||
| XM_017003745.1 | XP_016859234.1 | 12 - 13 | 1155 | 61 | ||
| GenBank mRNA | AK127836.1 | - | 1 - 2 | 1666 | 61 | |
| AK292952.1 | - | 11 - 12 | 1021 | 61 | ||
| AY039759.1 | - | 10 - 11 | 940 | 61 | ||
| BC012142.1 | - | 10 - 11 | 939 | 4647654 | 61 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000064.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
insulin receptor signaling pathway
ATP hydrolysis coupled proton transport
regulation of macroautophagy
positive regulation of Wnt signaling pathway
transferrin transport
ion transmembrane transport
phagosome acidification
hydrogen-exporting ATPase activity, phosphorylative mechanism
protein dimerization activity
insulin receptor signaling pathway
ATP hydrolysis coupled proton transport
regulation of macroautophagy
positive regulation of Wnt signaling pathway
transferrin transport
ion transmembrane transport
phagosome acidification
hydrogen-exporting ATPase activity, phosphorylative mechanism
protein dimerization activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | ENST00000272238.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | ENST00000272238.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | ENST00000272238.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | ENST00000272238.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | ENST00000272238.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | ENST00000272238.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | ENST00000272238.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | ENST00000272238.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | ENST00000272238.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | ENST00000272238.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | ENST00000272238.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | ENST00000272238.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | ENST00000272238.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | ENST00000272238.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | ENST00000272238.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | ENST00000381661.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | ENST00000381661.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | ENST00000381661.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | ENST00000381661.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | ENST00000381661.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | ENST00000381661.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | ENST00000381661.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | ENST00000381661.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | ENST00000381661.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | ENST00000381661.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | ENST00000381661.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | ENST00000381661.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | ENST00000381661.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | ENST00000381661.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | ENST00000381661.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | ENST00000480289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | ENST00000480289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | ENST00000480289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | ENST00000480289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | NM_001039362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | NM_001039362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | NM_001039362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | NM_001039362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | NM_001039362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | NM_001039362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | NM_001039362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | NM_001039362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | NM_001039362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | NM_001039362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | NM_001039362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | NM_001039362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | NM_001039362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | NM_001039362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | NM_001039362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | NM_001039362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | NM_144583.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | NM_144583.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | NM_144583.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | NM_144583.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | NM_144583.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | NM_144583.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | NM_144583.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | NM_144583.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | NM_144583.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | NM_144583.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | NM_144583.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | NM_144583.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | NM_144583.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | NM_144583.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006703.hg.1 | NM_144583.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02000824.hg.1 | ENST00000272238.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02000824.hg.1 | ENST00000381661.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02000824.hg.1 | NM_001039362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02000824.hg.1 | NM_144583.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02000824.hg.1 | TC02000064.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | ENST00000272238.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | ENST00000272238.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | ENST00000272238.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | ENST00000272238.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | ENST00000272238.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | ENST00000272238.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | ENST00000272238.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | ENST00000272238.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | ENST00000272238.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | ENST00000272238.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | ENST00000272238.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | ENST00000272238.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | ENST00000272238.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | ENST00000272238.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | ENST00000272238.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | ENST00000381661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | ENST00000381661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | ENST00000381661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | ENST00000381661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | ENST00000381661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | ENST00000381661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | ENST00000381661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | ENST00000381661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | ENST00000381661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | ENST00000381661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | ENST00000381661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | ENST00000381661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | ENST00000381661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | ENST00000381661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | ENST00000381661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | ENST00000480289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | ENST00000480289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | ENST00000480289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | ENST00000480289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | NM_001039362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | NM_001039362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | NM_001039362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | NM_001039362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | NM_001039362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | NM_001039362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | NM_001039362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | NM_001039362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | NM_001039362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | NM_001039362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | NM_001039362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | NM_001039362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | NM_001039362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | NM_001039362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | NM_001039362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | NM_001039362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | NM_144583.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | NM_144583.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | NM_144583.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | NM_144583.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | NM_144583.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | NM_144583.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | NM_144583.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | NM_144583.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | NM_144583.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | NM_144583.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | NM_144583.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | NM_144583.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | NM_144583.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | NM_144583.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006703.hg.1 | NM_144583.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200145967.hg.1 | ENST00000272238.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200145967.hg.1 | ENST00000381661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200145967.hg.1 | ENST00000480289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200145967.hg.1 | NM_001039362.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200145967.hg.1 | NM_144583.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200145967.hg.1 | ENST00000272238.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200145967.hg.1 | ENST00000381661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200145967.hg.1 | ENST00000480289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200145967.hg.1 | NM_001039362.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200145967.hg.1 | NM_144583.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200145967.hg.1 | TC0200006703.hg.1 | Clariom D TC |

ATP6V1C2
Genomic Map