Assay Details
| Gene Symbol: RBM41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171080.1 | NP_001164551.1 | 7 - 8 | 1260 | 104 | |
| NM_001324244.1 | NP_001311173.1 | 8 - 9 | 1332 | 104 | ||
| NR_136731.1 | 8 - 9 | 1126 | 104 | |||
| XM_011530980.2 | XP_011529282.1 | 8 - 9 | 1424 | 104 | ||
| GenBank mRNA | AK001878.1 | - | 7 - 8 | 1233 | 104 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X001259.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X016988.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X016988.hg.1 | TC0X001259.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000434854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000434854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000434854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000434854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000434854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000434854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000471079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000471079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000471079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000471079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000471079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000474499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000474499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000475556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000475556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000475556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000475556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000485676.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000485676.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000485676.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000485676.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000485676.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000485676.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000485676.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000485676.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X016989.hg.1 | ENST00000372487.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X016989.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X016989.hg.1 | TC0X001259.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000434854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000434854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000434854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000434854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000434854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000434854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000471079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000471079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000471079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000471079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000471079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000474499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000474499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000475556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000475556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000475556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000475556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000485676.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000485676.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000485676.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000485676.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000485676.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000485676.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000485676.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000485676.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | ENST00000495517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010462.hg.1 | NM_018301.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00171252.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00171252.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00171252.hg.1 | ENST00000372487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00171252.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00171252.hg.1 | TC0X00010462.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00171251.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00171251.hg.1 | NM_001171080.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00171251.hg.1 | TC0X00010462.hg.1 | Clariom D TC |

RBM41
Genomic Map