| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
ENST00000270257.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
ENST00000270257.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
ENST00000270257.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
ENST00000270257.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
ENST00000270257.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
ENST00000270257.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
ENST00000270257.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
ENST00000270257.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
ENST00000391951.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
ENST00000391951.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
ENST00000391951.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
ENST00000391951.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
ENST00000591607.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
ENST00000591607.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
ENST00000591607.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
ENST00000591607.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
ENST00000591607.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
ENST00000591747.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
ENST00000591747.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
ENST00000591747.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
ENST00000592351.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
ENST00000592351.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
ENST00000592351.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
NM_001007269.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
NM_001007269.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
NM_001007269.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
NM_001007269.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
NM_001007269.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
NM_001007269.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
NM_001007269.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
NM_001007270.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
NM_001007270.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
NM_001007270.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
NM_001007270.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
NM_001007270.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
NM_001007270.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
NM_001007270.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
NM_024707.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
NM_024707.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
NM_024707.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
NM_024707.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
NM_024707.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
NM_024707.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
NM_024707.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
NM_024707.2 |
Refseq |