Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19006520.hg.1 |
ENST00000246551.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19006520.hg.1 |
ENST00000437550.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19006520.hg.1 |
NM_001007469.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19006520.hg.1 |
NM_014266.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19006520.hg.1 |
TC19000489.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007891.hg.1 |
ENST00000246551.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007891.hg.1 |
ENST00000246551.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007891.hg.1 |
ENST00000246551.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007891.hg.1 |
ENST00000246551.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007891.hg.1 |
ENST00000246551.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007891.hg.1 |
ENST00000246551.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007891.hg.1 |
ENST00000246551.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007891.hg.1 |
ENST00000246551.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007891.hg.1 |
ENST00000437550.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007891.hg.1 |
ENST00000437550.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007891.hg.1 |
ENST00000437550.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007891.hg.1 |
ENST00000437550.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007891.hg.1 |
ENST00000437550.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007891.hg.1 |
ENST00000437550.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007891.hg.1 |
NM_001007469.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007891.hg.1 |
NM_001007469.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007891.hg.1 |
NM_001007469.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007891.hg.1 |
NM_001007469.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007891.hg.1 |
NM_001007469.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007891.hg.1 |
NM_001007469.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007891.hg.1 |
NM_001007469.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007891.hg.1 |
NM_014266.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007891.hg.1 |
NM_014266.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007891.hg.1 |
NM_014266.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007891.hg.1 |
NM_014266.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007891.hg.1 |
NM_014266.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007891.hg.1 |
NM_014266.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007891.hg.1 |
NM_014266.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900158802.hg.1 |
ENST00000246551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900158802.hg.1 |
ENST00000437550.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900158802.hg.1 |
NM_001007469.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900158802.hg.1 |
NM_014266.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900158802.hg.1 |
TC1900007891.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900158802.hg.1 |
ENST00000246551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900158802.hg.1 |
ENST00000437550.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900158802.hg.1 |
NM_001007469.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900158802.hg.1 |
NM_014266.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007891.hg.1 |
ENST00000246551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007891.hg.1 |
ENST00000246551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007891.hg.1 |
ENST00000246551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007891.hg.1 |
ENST00000246551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007891.hg.1 |
ENST00000246551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007891.hg.1 |
ENST00000246551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007891.hg.1 |
ENST00000246551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007891.hg.1 |
ENST00000246551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007891.hg.1 |
ENST00000437550.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007891.hg.1 |
ENST00000437550.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007891.hg.1 |
ENST00000437550.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007891.hg.1 |
ENST00000437550.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007891.hg.1 |
ENST00000437550.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007891.hg.1 |
ENST00000437550.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007891.hg.1 |
NM_001007469.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007891.hg.1 |
NM_001007469.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007891.hg.1 |
NM_001007469.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007891.hg.1 |
NM_001007469.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007891.hg.1 |
NM_001007469.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007891.hg.1 |
NM_001007469.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007891.hg.1 |
NM_001007469.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007891.hg.1 |
NM_014266.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007891.hg.1 |
NM_014266.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007891.hg.1 |
NM_014266.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007891.hg.1 |
NM_014266.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007891.hg.1 |
NM_014266.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007891.hg.1 |
NM_014266.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007891.hg.1 |
NM_014266.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900158801.hg.1 |
ENST00000246551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900158801.hg.1 |
ENST00000437550.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900158801.hg.1 |
NM_001007469.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900158801.hg.1 |
NM_014266.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900158801.hg.1 |
TC1900007891.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900158801.hg.1 |
ENST00000246551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900158801.hg.1 |
ENST00000437550.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900158801.hg.1 |
NM_001007469.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900158801.hg.1 |
NM_014266.3 |
Refseq |