| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
ENST00000450208.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030432.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030432.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030432.hg.1 |
TC01001960.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030433.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030433.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030433.hg.1 |
TC01001960.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030431.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030431.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030431.hg.1 |
TC01001960.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0100189335.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100189335.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100189335.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100189335.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100189335.hg.1 |
TC0100012123.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100189334.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100189334.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100189334.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100189334.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100189334.hg.1 |
TC0100012123.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
ENST00000450208.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100189333.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100189333.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100189333.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100189333.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100189333.hg.1 |
TC0100012123.hg.1 |
Clariom D TC |