| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
ENST00000450208.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030436.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030436.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01030436.hg.1 |
TC01001960.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0100189338.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100189338.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100189338.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100189338.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100189338.hg.1 |
TC0100012123.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
ENST00000450208.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |