Assay Details
| Gene Symbol: IGSF21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032880.4 | NP_116269.3 | 1 - 2 | 456 | 59 | |
| XM_011542319.2 | XP_011540621.1 | 1 - 2 | 413 | 59 | ||
| GenBank mRNA | AK075316.1 | - | 1 - 2 | 456 | 59 | |
| BC007618.1 | - | 1 - 2 | 376 | 3355289 | 59 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000239.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01003812.hg.1 | ENST00000251296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01003812.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01003812.hg.1 | TC01000239.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000412684.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000412684.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000412684.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000412684.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000412684.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000412684.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000473951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000473951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000473951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000473951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000497331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000497331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000497331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000497331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000497331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000497331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000497331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000497331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000497331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | ENST00000497331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01003811.hg.1 | ENST00000251296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01003811.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01003811.hg.1 | TC01000239.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01003813.hg.1 | ENST00000251296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01003813.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01003813.hg.1 | TC01000239.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0100149754.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100149754.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100149754.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100149754.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100149754.hg.1 | TC0100007136.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100149752.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100149752.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100149752.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100149752.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100149752.hg.1 | TC0100007136.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100149753.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100149753.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100149753.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100149753.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100149753.hg.1 | TC0100007136.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100149755.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100149755.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100149755.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100149755.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100149755.hg.1 | TC0100007136.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000251296.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000412684.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000412684.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000412684.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000412684.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000412684.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000412684.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000473951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000473951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000473951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000473951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000497331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000497331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000497331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000497331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000497331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000497331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000497331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000497331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000497331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | ENST00000497331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | NM_032880.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007136.hg.1 | NM_032880.4 | Refseq |

IGSF21
Genomic Map