| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
ENST00000361531.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
ENST00000361531.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
ENST00000361531.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
ENST00000361531.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
ENST00000361531.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
ENST00000361531.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
ENST00000361531.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
ENST00000361531.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
ENST00000361531.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
ENST00000368211.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
ENST00000368211.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
ENST00000368211.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
ENST00000368211.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
ENST00000368211.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
ENST00000368211.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
ENST00000368211.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
ENST00000368211.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
ENST00000368211.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
ENST00000412846.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
ENST00000412846.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
ENST00000412846.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
ENST00000412846.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
ENST00000412846.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
ENST00000412846.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
ENST00000420938.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
ENST00000420938.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
ENST00000420938.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
ENST00000434558.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
ENST00000434558.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
ENST00000434558.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
ENST00000434558.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
ENST00000434558.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
ENST00000434558.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
ENST00000478899.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
ENST00000478899.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
ENST00000478899.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
ENST00000478899.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
ENST00000478899.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
NM_024540.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
NM_024540.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
NM_024540.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
NM_024540.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
NM_024540.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
NM_024540.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
NM_024540.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
NM_024540.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
NM_024540.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
NM_145729.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
NM_145729.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
NM_145729.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
NM_145729.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
NM_145729.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
NM_145729.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
NM_145729.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
NM_145729.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
NM_145729.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016021.hg.1 |
NM_145729.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052344.hg.1 |
ENST00000361531.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052344.hg.1 |
ENST00000368211.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052344.hg.1 |
ENST00000478899.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052344.hg.1 |
NM_024540.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052344.hg.1 |
NM_145729.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01052344.hg.1 |
TC01003352.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
ENST00000361531.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
ENST00000361531.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
ENST00000361531.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
ENST00000361531.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
ENST00000361531.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
ENST00000361531.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
ENST00000361531.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
ENST00000361531.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
ENST00000361531.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
ENST00000368211.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
ENST00000368211.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
ENST00000368211.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
ENST00000368211.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
ENST00000368211.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
ENST00000368211.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
ENST00000368211.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
ENST00000368211.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
ENST00000368211.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
ENST00000412846.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
ENST00000412846.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
ENST00000412846.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
ENST00000412846.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
ENST00000412846.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
ENST00000412846.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
ENST00000420938.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
ENST00000420938.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
ENST00000420938.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
ENST00000434558.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
ENST00000434558.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
ENST00000434558.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
ENST00000434558.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
ENST00000434558.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
ENST00000434558.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
ENST00000478899.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
ENST00000478899.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
ENST00000478899.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
ENST00000478899.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
ENST00000478899.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
NM_024540.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
NM_024540.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
NM_024540.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
NM_024540.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
NM_024540.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
NM_024540.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
NM_024540.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
NM_024540.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
NM_024540.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
NM_145729.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
NM_145729.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
NM_145729.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
NM_145729.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
NM_145729.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
NM_145729.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
NM_145729.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
NM_145729.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
NM_145729.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016021.hg.1 |
NM_145729.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100221566.hg.1 |
ENST00000361531.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100221566.hg.1 |
ENST00000368211.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100221566.hg.1 |
ENST00000434558.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100221566.hg.1 |
ENST00000478899.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100221566.hg.1 |
NM_024540.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100221566.hg.1 |
NM_145729.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100221566.hg.1 |
ENST00000361531.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100221566.hg.1 |
ENST00000368211.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100221566.hg.1 |
ENST00000434558.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100221566.hg.1 |
ENST00000478899.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100221566.hg.1 |
NM_024540.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100221566.hg.1 |
NM_145729.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100221566.hg.1 |
TC0100016021.hg.1 |
Clariom D TC |