Assay Details
| Gene Symbol: ADGRG7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001308362.1 | NP_001295291.1 | 6 - 7 | 1171 | 82 | |
| NM_032787.2 | NP_116176.2 | 12 - 13 | 1995 | 82 | ||
| XM_011513245.2 | XP_011511547.1 | 11 - 12 | 1870 | 82 | ||
| GenBank mRNA | AK027360.1 | - | 12 - 13 | 1983 | 82 | |
| AK131069.1 | - | 5 - 6 | 854 | 82 | ||
| AK172769.1 | - | 8 - 9 | 1237 | 82 | ||
| AK172774.1 | - | 4 - 5 | 748 | 82 | ||
| AK295345.1 | - | 6 - 7 | 1113 | 82 | ||
| AY181246.1 | - | 12 - 13 | 1727 | 82 | ||
| BC113451.1 | - | 12 - 13 | 1761 | 8322503 | 82 | |
| BC130388.1 | - | 12 - 13 | 1761 | 40146419 | 82 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000510.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000481361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000481361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000481361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000481361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000481361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000481361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000481506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000481506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000481506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000481506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000481506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000481506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000481506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000481506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000481506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000493081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000493081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000493081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000493081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000493081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009612.hg.1 | ENST00000273352.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009612.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009612.hg.1 | ENST00000481506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009612.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009612.hg.1 | TC03000510.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000481361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000481361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000481361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000481361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000481361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000481361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000481506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000481506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000481506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000481506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000481506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000481506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000481506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000481506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000481506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000493081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000493081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000493081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000493081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000493081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300158111.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158111.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158111.hg.1 | ENST00000481506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158111.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300158111.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300158111.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158111.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158111.hg.1 | ENST00000481506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158111.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300158111.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300158111.hg.1 | TC0300008150.hg.1 | Clariom D TC |

ADGRG7
Genomic Map