Assay Details
| Gene Symbol: ADGRG7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032787.2 | NP_116176.2 | 8 - 9 | 1218 | 160 | |
| XM_011513245.2 | XP_011511547.1 | 7 - 8 | 1093 | 160 | ||
| GenBank mRNA | AK027360.1 | - | 8 - 9 | 1206 | 160 | |
| AK131069.1 | - | 1 - 2 | 77 | 160 | ||
| AK172769.1 | - | 4 - 5 | 460 | 160 | ||
| AY181246.1 | - | 8 - 9 | 950 | 160 | ||
| BC113451.1 | - | 8 - 9 | 984 | 8322503 | 160 | |
| BC130388.1 | - | 8 - 9 | 984 | 40146419 | 160 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000510.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000481361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000481361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000481361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000481361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000481361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000481361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000481506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000481506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000481506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000481506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000481506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000481506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000481506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000481506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000481506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000493081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000493081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000493081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000493081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | ENST00000493081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009607.hg.1 | ENST00000273352.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009607.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009607.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009607.hg.1 | TC03000510.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0300158105.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158105.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300158105.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158105.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300158105.hg.1 | TC0300008150.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000481361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000481361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000481361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000481361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000481361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000481361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000481506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000481506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000481506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000481506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000481506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000481506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000481506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000481506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000481506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000493081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000493081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000493081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000493081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | ENST00000493081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008150.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300158106.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158106.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158106.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300158106.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300158106.hg.1 | ENST00000273352.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158106.hg.1 | ENST00000475887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158106.hg.1 | NM_001308362.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300158106.hg.1 | NM_032787.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300158106.hg.1 | TC0300008150.hg.1 | Clariom D TC |

ADGRG7
Genomic Map