Assay Details
| Gene Symbol: MUC13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033049.3 | NP_149038.3 | 3 - 4 | 681 | 154 | |
| GenBank mRNA | AB035807.1 | - | 3 - 4 | 666 | 154 | |
| AF286113.1 | - | 3 - 4 | 669 | 154 | ||
| AY358831.1 | - | 3 - 4 | 666 | 154 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001719.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | ENST00000462728.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | ENST00000462728.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | ENST00000462728.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | ENST00000478191.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | ENST00000478191.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | ENST00000478191.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | ENST00000478191.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | ENST00000478191.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | ENST00000478191.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | ENST00000478191.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | ENST00000490147.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | ENST00000490147.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | ENST00000490147.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | ENST00000490147.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | ENST00000490147.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | ENST00000490147.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | ENST00000497378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | ENST00000497378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | ENST00000497378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | ENST00000497378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | ENST00000497378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | ENST00000616727.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | ENST00000616727.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | ENST00000616727.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | ENST00000616727.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | ENST00000616727.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | ENST00000616727.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | ENST00000616727.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | ENST00000616727.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | ENST00000616727.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | ENST00000616727.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | ENST00000616727.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | ENST00000616727.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | ENST00000616727.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | ENST00000616727.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | ENST00000616727.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | ENST00000616727.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | ENST00000616727.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | NM_033049.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | NM_033049.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | NM_033049.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | NM_033049.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | NM_033049.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | NM_033049.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | NM_033049.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | NM_033049.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | NM_033049.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | NM_033049.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | NM_033049.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | NM_033049.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | NM_033049.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | NM_033049.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | NM_033049.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | NM_033049.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012236.hg.1 | NM_033049.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03031262.hg.1 | ENST00000311075.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03031262.hg.1 | NM_033049.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03031262.hg.1 | TC03001719.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0300191325.hg.1 | ENST00000478191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300191325.hg.1 | ENST00000490147.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300191325.hg.1 | ENST00000616727.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300191325.hg.1 | NM_033049.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300191325.hg.1 | ENST00000478191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300191325.hg.1 | ENST00000490147.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300191325.hg.1 | ENST00000616727.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300191325.hg.1 | NM_033049.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300191325.hg.1 | TC0300012236.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | ENST00000462728.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | ENST00000462728.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | ENST00000462728.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | ENST00000478191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | ENST00000478191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | ENST00000478191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | ENST00000478191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | ENST00000478191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | ENST00000478191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | ENST00000478191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | ENST00000490147.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | ENST00000490147.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | ENST00000490147.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | ENST00000490147.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | ENST00000490147.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | ENST00000490147.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | ENST00000497378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | ENST00000497378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | ENST00000497378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | ENST00000497378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | ENST00000497378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | ENST00000616727.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | ENST00000616727.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | ENST00000616727.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | ENST00000616727.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | ENST00000616727.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | ENST00000616727.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | ENST00000616727.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | ENST00000616727.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | ENST00000616727.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | ENST00000616727.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | ENST00000616727.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | ENST00000616727.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | ENST00000616727.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | ENST00000616727.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | ENST00000616727.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | ENST00000616727.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | ENST00000616727.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | NM_033049.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | NM_033049.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | NM_033049.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | NM_033049.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | NM_033049.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | NM_033049.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | NM_033049.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | NM_033049.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | NM_033049.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | NM_033049.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | NM_033049.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | NM_033049.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | NM_033049.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | NM_033049.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | NM_033049.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | NM_033049.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012236.hg.1 | NM_033049.3 | Refseq |

MUC13
Genomic Map