Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC100507091 | LOC100507091 | -2.7kb | Anti-sense | Chr. 12 - 123081384 - 123084744 on NCBI Build GRCh38 | |
| Downstream | M-phase phosphoprotein 9 | MPHOSPH9 |
MPP-9, MPP9 |
59.8kb | Sense | Chr. 12 - 123152324 - 123244014 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map