| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012129.hg.1 |
ENST00000306108.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012129.hg.1 |
ENST00000306108.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012129.hg.1 |
ENST00000306108.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012129.hg.1 |
ENST00000306108.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012129.hg.1 |
ENST00000306108.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012129.hg.1 |
ENST00000306108.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012129.hg.1 |
ENST00000306108.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012129.hg.1 |
ENST00000306108.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012129.hg.1 |
ENST00000306108.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012129.hg.1 |
ENST00000306108.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012129.hg.1 |
ENST00000419999.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012129.hg.1 |
ENST00000419999.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012129.hg.1 |
ENST00000419999.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012129.hg.1 |
ENST00000419999.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012129.hg.1 |
ENST00000419999.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012129.hg.1 |
ENST00000439947.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012129.hg.1 |
ENST00000439947.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012129.hg.1 |
ENST00000439947.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012129.hg.1 |
ENST00000439947.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012129.hg.1 |
ENST00000439947.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012129.hg.1 |
ENST00000439947.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012129.hg.1 |
ENST00000439947.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012129.hg.1 |
ENST00000484060.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012129.hg.1 |
ENST00000484060.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012129.hg.1 |
ENST00000484060.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012129.hg.1 |
ENST00000490390.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012129.hg.1 |
ENST00000490390.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012129.hg.1 |
ENST00000490390.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012129.hg.1 |
ENST00000490390.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012129.hg.1 |
NM_017910.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012129.hg.1 |
NM_017910.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012129.hg.1 |
NM_017910.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012129.hg.1 |
NM_017910.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012129.hg.1 |
NM_017910.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012129.hg.1 |
NM_017910.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012129.hg.1 |
NM_017910.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012129.hg.1 |
NM_017910.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012129.hg.1 |
NM_017910.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012129.hg.1 |
NM_017910.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02027453.hg.1 |
ENST00000306108.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02027453.hg.1 |
ENST00000484060.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02027453.hg.1 |
NM_017910.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02027453.hg.1 |
TC02001703.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0200188530.hg.1 |
ENST00000306108.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200188530.hg.1 |
ENST00000439947.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200188530.hg.1 |
ENST00000484060.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200188530.hg.1 |
ENST00000490390.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200188530.hg.1 |
NM_017910.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200188530.hg.1 |
ENST00000306108.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200188530.hg.1 |
ENST00000439947.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200188530.hg.1 |
ENST00000484060.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200188530.hg.1 |
ENST00000490390.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200188530.hg.1 |
NM_017910.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200188530.hg.1 |
TC0200012129.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200012129.hg.1 |
ENST00000306108.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012129.hg.1 |
ENST00000306108.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012129.hg.1 |
ENST00000306108.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012129.hg.1 |
ENST00000306108.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012129.hg.1 |
ENST00000306108.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012129.hg.1 |
ENST00000306108.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012129.hg.1 |
ENST00000306108.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012129.hg.1 |
ENST00000306108.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012129.hg.1 |
ENST00000306108.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012129.hg.1 |
ENST00000306108.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012129.hg.1 |
ENST00000419999.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012129.hg.1 |
ENST00000419999.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012129.hg.1 |
ENST00000419999.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012129.hg.1 |
ENST00000419999.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012129.hg.1 |
ENST00000419999.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012129.hg.1 |
ENST00000439947.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012129.hg.1 |
ENST00000439947.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012129.hg.1 |
ENST00000439947.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012129.hg.1 |
ENST00000439947.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012129.hg.1 |
ENST00000439947.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012129.hg.1 |
ENST00000439947.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012129.hg.1 |
ENST00000439947.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012129.hg.1 |
ENST00000484060.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012129.hg.1 |
ENST00000484060.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012129.hg.1 |
ENST00000484060.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012129.hg.1 |
ENST00000490390.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012129.hg.1 |
ENST00000490390.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012129.hg.1 |
ENST00000490390.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012129.hg.1 |
ENST00000490390.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012129.hg.1 |
NM_017910.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012129.hg.1 |
NM_017910.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012129.hg.1 |
NM_017910.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012129.hg.1 |
NM_017910.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012129.hg.1 |
NM_017910.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012129.hg.1 |
NM_017910.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012129.hg.1 |
NM_017910.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012129.hg.1 |
NM_017910.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012129.hg.1 |
NM_017910.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012129.hg.1 |
NM_017910.3 |
Refseq |